Skip to Content
Merck
All Photos(2)

Key Documents

AB19013

Sigma-Aldrich

Anti-Tenascin Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-150-225, Anti-DFNA56, Anti-GMEM, Anti-GP, Anti-HXB, Anti-JI, Anti-TN, Anti-TN-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

chicken, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

Specificity

Reactive with tenascin from chicken and human by immunohistochemistry. Under reducing conditions, reactivity to chicken is approximately 100X greater than reactivity with human tenascin. Reactivity with mouse tenascin under reducing conditions is poor, but reacts with mouse in paraffin embedded sections.

Immunogen

Purified tenascin from embryonic chicken brain.

Application

Anti-Tenascin Antibody is an antibody against Tenascin for use in IH, IH(P).
Function blocking

Western blot: Antibody recognizes a 220-240kDa band in reduced westerns; antibody reacts best with chicken, followed by human (100X less reactive). Mouse reactivity under reduced conditions is poor.

Immunohistochemistry: frozen and paraffin embedded tissues; citrate treatment useful for paraffin tissues.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Linkage

Replaces: AB1951

Physical form

Affinity Purified using immobilized chicken tenascin. Provided as a liquid in PBS, containing no preservatives.
ImmunoAffinity Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Embryonic chicken brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent whole-mount method for visualizing three-dimensional relationships in intact and regenerating adult newt spinal cords.
Zukor, KA; Kent, DT; Odelberg, SJ
Developmental Dynamics null
The impact of flow-induced forces on the morphogenesis of the outflow tract.
Biechler, SV; Junor, L; Evans, AN; Eberth, JF; Price, RL; Potts, JD; Yost, MJ; Goodwin, RL
Frontiers in Physiology null
Ashley W Seifert et al.
PloS one, 7(4), e32875-e32875 (2012-04-10)
While considerable progress has been made towards understanding the complex processes and pathways that regulate human wound healing, regenerative medicine has been unable to develop therapies that coax the natural wound environment to heal scar-free. The inability to induce perfect
Guoqiang Wan et al.
Frontiers in aging neuroscience, 11, 156-156 (2019-07-12)
Age-related decline of inner ear function contributes to both hearing loss and balance disorders, which lead to impaired quality of life and falls that can result in injury and even death. The cellular mechanisms responsible for the ear's functional decline
Hong Tan et al.
Developmental biology, 374(2), 345-356 (2012-12-25)
Fibrous development of the extracellular matrix (ECM) of cardiac valves is necessary for proper heart function. Pathological remodeling of valve ECM is observed in both pediatric and adult cardiac disorders. It is well established that intracardiac hemodynamics play a significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service