Skip to Content
Merck
All Photos(1)

Key Documents

AB1891

Sigma-Aldrich

Anti-C-X-X-X-C Chemokine Receptor 1 Antibody, extracellular loop

Chemicon®, from rabbit

Synonym(s):

CX3CR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CX3CR1(1524)

General description

CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, mouse, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively (1-4). The seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1 (5). Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by facktokine (6). CX3CR1 mediates leukocyte migration and adhesion.

Immunogen

Synthetic peptide from the extracellular loop of human CX3CR1.

Application

Anti-C-X-X-X-C Chemokine Receptor 1 Antibody, extracellular loop is an antibody against C-X-X-X-C Chemokine Receptor 1 for use in IH & WB.
Western blot: 1:500 (THP cell lysate can be used as positive control and an approximately 50 kDa band can be detected)

Immunohistochemistry in frozen sections

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Kajiwara et al.
Journal of immunology (Baltimore, Md. : 1950), 193(1), 335-343 (2014-06-01)
Response to endotoxins is an important part of the organismal reaction to Gram-negative bacteria and plays a critical role in sepsis and septic shock, as well as other conditions such as metabolic endotoxemia. Humans are generally more sensitive to endotoxins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service