Skip to Content
Merck
All Photos(1)

Key Documents

AB1555

Sigma-Aldrich

Anti-NMDAR2A Antibody

serum, Chemicon®

Synonym(s):

Anti-EPND, Anti-GluN2A, Anti-LKS, Anti-NMDAR2A, Anti-NR2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

fish, human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

suitability

not suitable for immunohistochemistry

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

Specificity

NMDAR2A. By Western blot it recognizes a 180 kDa band in rat brain membranes.

Immunogen

C-Terminal Fusion Protein of the NMDAR2A (30 kDa), aa 1253-1391.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-NMDAR2A Antibody is validated for use in IP, WB for the detection of NMDAR2A.
Western blotting*: 1:1,000-1:5,000 for ECL. If using labled protein-A for detection: 1:200.

* See suggested protocol

Immunoprecipitation using solubilized hippocampal slices. 3μL will (under appropriate conditions) quantitatively immunoprecipitate all NMDAR2A in 200μg rat brain.

Not suggested for use in immunohistochemistry.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit antiserum. Liquid, no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to one year. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
NMDAR2A is present in high concentrations in the hippocampus

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen-Zhen Duan et al.
Scientific reports, 6, 29246-29246 (2016-07-08)
It has been demonstrated that Src could modulate NMDA receptor, and PAR1 could also affect NMDAR signaling. However, whether PAR1 could regulate NMDAR through Src under ICH has not yet been investigated. In this study, we demonstrated the role of
Annabelle Schlüter et al.
Molecular therapy. Methods & clinical development, 17, 281-299 (2020-02-15)
In the central nervous system, neurons and the vasculature influence each other. While it is well described that a functional vascular system is trophic to neurons and that vascular damage contributes to neurodegeneration, the opposite scenario in which neural damage
Sharon A Swanger et al.
American journal of human genetics, 99(6), 1261-1280 (2016-11-15)
Epilepsy and intellectual disability are associated with rare variants in the GluN2A and GluN2B (encoded by GRIN2A and GRIN2B) subunits of the N-methyl-D-aspartate receptor (NMDAR), a ligand-gated ion channel with essential roles in brain development and function. By assessing genetic
Effects of repeated prenatal glucocorticoid exposure on long-term potentiation in the juvenile guinea-pig hippocampus.
Setiawan, E; Jackson, MF; MacDonald, JF; Matthews, SG
The Journal of Physiology null
Visual experience-independent functional expression of NMDA receptors in the developing rabbit retina.
Chang, YC; Chen, CY; Chiao, CC
Investigative Ophthalmology & Visual Science null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service