Skip to Content
Merck
All Photos(1)

Documents

AB15360

Sigma-Aldrich

Anti-Calcitonin Gene Related Peptide Antibody

serum, Chemicon®

Synonym(s):

CGRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CALCA(796)

Specificity

Recognizes Calcitonin Gene Related Peptide (CGRP).

Immunogen

Synthetic calcitonin gene related peptide.

Application

Anti-Calcitonin Gene Related Peptide Antibody detects level of Calcitonin Gene Related Peptide & has been published & validated for use in IH.
Immunohistochemistry on frozen rat spinal cord: 1:2,000-1:4,000 using the Chemicon IHC-Select Kit with HRP-DAB detection system.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Linkage

Replaces: AB1971

Physical form

Rabbit serum. Liquid.
Serum

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea M Sartori et al.
Experimental neurology, 348, 113937-113937 (2021-11-27)
Neurogenic lower urinary tract dysfunction typically develops after spinal cord injury. We investigated the time course and the anatomical changes in the spinal cord that may be causing lower urinary tract symptoms following injury. Rats were implanted with a bladder
Andrew H Cooper et al.
Pain reports, 5(6), e872-e872 (2020-12-05)
Inflammation during the neonatal period can exacerbate pain severity following reinjury in adulthood. This is driven by alterations in the postnatal development of spinal and supraspinal nociceptive circuitry. However, the contribution of alterations in peripheral nociceptor function remains underexplored. We
A virus-like particle-based anti-nerve growth factor vaccine reduces inflammatory hyperalgesia: potential long-term therapy for chronic pain.
Rohn, TA; Ralvenius, WT; Paul, J; Borter, P; Hernandez, M; Witschi, R; Grest et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Michał Bulc et al.
Journal of diabetes research, 2018, 4735659-4735659 (2018-08-25)
One of the most frequently reported disorders associated with diabetes is gastrointestinal (GI) disturbance. Although pathogenesis of these complications is multifactorial, the complicity of the enteric nervous system (ENS) in this respect has significant importance. Therefore, this paper analysed changes
Devin M Barry et al.
Molecular pain, 12 (2016-04-14)
There are substantial disagreements about the expression of gastrin-releasing peptide (GRP) in sensory neurons and whether GRP antibody cross-reacts with substance P (SP). These concerns necessitate a critical revaluation of GRP expression using additional approaches. Here, we show that a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service