8.55087
FMPB NovaGel HL
Novabiochem®
Synonym(s):
4-(4-Formyl-3-methoxyphenoxy)butyryl NovaGel HL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111023
NACRES:
NA.22
Recommended Products
Quality Level
product line
NovaGel
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
aldehyde
storage temp.
2-8°C
General description
This product is prepared from aminomethyl NovaGel by derivatization with the 4-(4-formyl-3-methoxyphenoxy)butanoic acid. For further information on NovaGel supports, please refer to the product entry for 855084.
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Linkage
Replaces: 01-64-0287
Analysis Note
Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis of sulfur after derivatization with tosylhydrazide): 0.30 - 0.70 mmol/g
Swelling Volume (in DMF): lot specific result
The PEG PS resin is prepared from aminoethyl copoly (styrene- 1% DVB) , 100 - 200 mesh
Appearance of substance (visual): beads
Loading (determined by elemental analysis of sulfur after derivatization with tosylhydrazide): 0.30 - 0.70 mmol/g
Swelling Volume (in DMF): lot specific result
The PEG PS resin is prepared from aminoethyl copoly (styrene- 1% DVB) , 100 - 200 mesh
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service