8.51012
HCTU
O-(1H-6-Chlorobenzotriazole-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate Novabiochem®
Synonym(s):
2-(6-Chloro-1-H-benzotriazole-1-yl)-1,1,3,3-tetramethylaminium hexafluorophosphate, HCTU
About This Item
Quality Level
product line
Novabiochem®
Assay
≥98% (HPLC)
form
powder
reaction suitability
reaction type: Coupling Reactions
manufacturer/tradename
Novabiochem®
mp
>185 °C
application(s)
peptide synthesis
storage temp.
2-8°C
InChI
1S/C11H15ClN5O.F6P/c1-15(2)11(16(3)4)18-17-10-7-8(12)5-6-9(10)13-14-17;1-7(2,3,4,5)6/h5-7H,1-4H3;/q+1;-1
InChI key
ZHHGTMQHUWDEJF-UHFFFAOYSA-N
General description
Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents
Literature references
[1] O. Marder, et al. (2002) Chimica Oggi, 37.
[2] G. Sabatino, et al. in ′Peptides 2002, Proceedings of the 27th European Peptide Symposium′, E. Benedetti & C. Pedone (Eds), Naples, Edizioni Ziino, 2002, pp. 272.
[3] M. Gude & S. Barthélémy in ′Peptides 2002, Proceedings of the 27th European Peptide Symposium′, E. Benedetti & C. Pedone (Eds), Naples, Edizioni Ziino, 2002, pp. 122.
[4] G. Sabatino, et al. in ′Peptide Revolution: Genomics, Proteomics & Therapeutics, , Proc. 18th American Peptide Symposium′, M. Chrev & T. K. Sawyer (Eds), Cardiff, American Peptide Society, 2003, pp. 49.
Application
- High-throughput parallel synthesis optimization of Glucagon-like Peptide 1 receptor agonists: Describes the use of HCTU as a reactive coupling reagent in peptide synthesis, highlighting its effectiveness in the production of GLP-1 agonists. (Ramos-Colón et al., 2018).
- An efficient synthesis of quinoxaline derivatives using HCTU as catalyst in DMF: Outlines a novel method for synthesizing quinoxaline and its derivatives using HCTU, highlighting its catalytic properties in facilitating efficient chemical reactions. (Sasane et al., 2023).
- An efficient one-pot conversion of carboxylic acids into benzimidazoles via an HBTU-promoted methodology: Describes the use of HCTU in a methodology to convert carboxylic acids into benzimidazoles, noting its role alongside other carbodiimide agents. (Barasa and Yoganathan, 2018).
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Water (K. F.): ≤ 0.50 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1A
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service