Skip to Content
Merck
All Photos(2)

Key Documents

8.43947

Sigma-Aldrich

Ethylene glycol monopropyl ether

for synthesis

Synonym(s):

2-Propoxyethanol, Propyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1731983
MDL number:
UNSPSC Code:
12352112
EC Index Number:
220-548-6
NACRES:
NA.22
grade:
for synthesis
bp:
150-153 °C (lit.)
vapor pressure:
1.3 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

1.3 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

230 °C

potency

3089 mg/kg LD50, oral (Rat)
876 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

refractive index

n20/D 1.413 (lit.)

bp

150-153 °C (lit.)

mp

−75 °C (lit.)

transition temp

flash point 48 °C

density

0.913 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCOCCO

InChI

1S/C5H12O2/c1-2-4-7-5-3-6/h6H,2-5H2,1H3

InChI key

YEYKMVJDLWJFOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.897 - 0.900
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test
Evaporation Number: 68

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service