Skip to Content
Merck
All Photos(5)

Key Documents

8.22271

Supelco

Dichloromethane

EMPLURA®

Synonym(s):

DCM, Methylene chloride, Methylene dichloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-838-9
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

1223 °F
605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

22 %

impurities

≤0.1% Water

evapn. residue

≤0.0020%

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

solubility

20 g/L

density

1.325 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 1.323 - 1.326
Evaporation residue: ≤ 0.0020 %
Water: ≤ 0.10 %
Stabilized with 2-Methyl-2-buten

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service