Skip to Content
Merck
All Photos(1)

Key Documents

8.20175

Sigma-Aldrich

2-Bromoethanol

for synthesis

Synonym(s):

2-Bromoethanol, Ethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
124.97
MDL number:
UNSPSC Code:
12352104
EC Index Number:
208-748-1
NACRES:
NA.22
form:
liquid

vapor pressure

3.2 hPa ( 20 °C)

Quality Level

form

liquid

bp

56-57 °C/27 hPa

mp

-80 °C

transition temp

flash point 40 °C

density

1.76 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCCO

InChI

1S/C2H5BrO/c3-1-2-4/h4H,1-2H2

InChI key

LDLCZOVUSADOIV-UHFFFAOYSA-N

General description

for synthesis

Application

2-Bromoethanol for synthesis. CAS 540-51-2, EC Number 208-748-1, molar mass 124.97 g/mol.

Analysis Note

Assay (GC, area%)≥ 97.0 % (a/a)Density (d 20 °C/ 4 °C)1.760 - 1.765Identity (IR)passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service