Skip to Content
Merck
All Photos(1)

Key Documents

8.18897

Sigma-Aldrich

Polyethylene glycol

20000 (stabilised) for synthesis

Synonym(s):

Poly(ethylene glycol), PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

flakes

autoignition temp.

>320 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

58-63 °C

transition temp

flash point 240 °C

solubility

500 g/L

density

1.2 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 20000 (PEG-20000) can be used as a pore-expanding agent to prepare mesoporous La2O3 catalyst via hydrothermal process. The effect of PEG 20000 on La2O3 catalyst particles, results in increased alkalinity, crystallinity, specific surface area, and dispersion. These modifications of the catalyst improve the catalytic performance in the transesterification of dimethyl carbonate with glycerol.[1]
It can also be used as:
  • A surfactant in the synthesis of nanomaterials by various methods to control the size and morphology of nanostructure.[2][3]
  • A support to prepare zero-valent iron-PEG nanocomposite material (Fe/PEG) applicable in the removal of Pb(II) from aqueous solutions.[4]

Analysis Note

Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 66 °C
Hydroxyl value: 4 - 7
Average molecular mass: 16000 - 28000

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monodisperse mesoporous La2O3 flakes for the synthesis of glycerol carbonate by efficiently catalyzing the transesterification of dimethyl carbonate with glycerol
Yue Chengguang, et al.
Reaction Kinetics, Mechanisms and Catalysis, 128(2), 763-778 (2019)
Polyethylene Glycol 20000-Supported Zero-Valent Iron Nanoparticle for Removal of Pb (II) from Aqueous Solutions: Characteristics, Kinetic and Thermodynamic Studies
He Qi, et al.
Science of Advanced Materials, 8(10), 1878-1886 (2016)
Hydrothermal synthesis of hexagonal magnesium hydroxide nanoflakes
Wang Qiang, et al.
Materials Research Bulletin, 51, 35-39 (2014)
Microwave-assisted rapid synthesis of γ-cyclodextrin metal-organic frameworks for size control and efficient drug loading
Liu Botao, et al.
Crystal Growth & Design, 17(4), 1654-1660 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service