Skip to Content
Merck
All Photos(1)

Key Documents

8.08146

Sigma-Aldrich

1-Tetradecanol

for synthesis

Synonym(s):

1-Tetradecanol, Myristyl alcohol, Tetradecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H30O
CAS Number:
Molecular Weight:
214.39
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-000-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

240 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

277-288 °C/1013 hPa

mp

36-39 °C

transition temp

flash point 145 °C

density

0.8 g/cm3 at 60 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCCCC

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3

InChI key

HLZKNKRTKFSKGZ-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

293.0 °F

Flash Point(C)

145 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service