Skip to Content
Merck
All Photos(1)

Key Documents

8.04744

Sigma-Aldrich

2-Iodoacetamide

for synthesis

Synonym(s):

2-Iodoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4INO
CAS Number:
Molecular Weight:
184.96
MDL number:
UNSPSC Code:
12352111
EC Index Number:
205-630-1
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

91-94 °C

storage temp.

2-8°C

SMILES string

ICC(=O)N

InChI

1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

PGLTVOMIXTUURA-UHFFFAOYSA-N

Analysis Note

Purity (HPLC, area%): ≥ 99
Assay (ex N): ≥ 98
Identity (IR-spectrum): passes test
Colour (2.5 %, water): ≤ 50 APHA
Melting point: >91
Fe (Iron): ≤ 20 ppm
Water (according to Karl Fischer): ≤ 3

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service