Skip to Content
Merck
All Photos(1)

Key Documents

70954

Sigma-Aldrich

Rosetta(DE3) Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

BL21 derivatives

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

Product Name

Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

shipped in

wet ice

storage temp.

−70°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm (DE3) pRARE (CamR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta strains provide for “universal” translation which is otherwise limited by the codon usage of E. coli. The tRNA genes are driven by their native promoters. In Rosetta(DE3)pLysS, the rare tRNA genes are present on the same plasmids that carries the T7 lysozyme gene.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta(DE3) Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suman Rao et al.
Cell chemical biology, 26(6), 818-829 (2019-04-16)
Covalent kinase inhibitors, which typically target cysteine residues, represent an important class of clinically relevant compounds. Approximately 215 kinases are known to have potentially targetable cysteines distributed across 18 spatially distinct locations proximal to the ATP-binding pocket. However, only 40
Gilberto Betancor et al.
Cell reports, 29(7), 1923-1933 (2019-11-14)
Myxovirus resistance 2 (MX2/MXB) is an interferon (IFN)-induced HIV-1 restriction factor that inhibits viral nuclear DNA accumulation. The amino-terminal domain of MX2 binds the viral capsid and is essential for inhibition. Using in vitro assembled Capsid-Nucleocapsid (CANC) complexes as a surrogate
Moonsup Lee et al.
The Journal of cell biology, 218(8), 2659-2676 (2019-07-05)
Cilia are critical for proper embryonic development and maintaining homeostasis. Although extensively studied, there are still significant gaps regarding the proteins involved in regulating ciliogenesis. Using the Xenopus laevis embryo, we show that Dishevelled (Dvl), a key Wnt signaling scaffold
Markus Hassler et al.
Molecular cell, 74(6), 1175-1188 (2019-06-22)
The condensin protein complex plays a key role in the structural organization of genomes. How the ATPase activity of its SMC subunits drives large-scale changes in chromosome topology has remained unknown. Here we reconstruct, at near-atomic resolution, the sequence of
Michael Lisby et al.
STAR protocols, 1(2) (2020-09-29)
Human ZGRF1 is one of the many helicases that play important roles in the repair of damaged chromosomes by homologous recombination. However, the lack of biochemical characterization of the ZGRF1 protein has hindered the understanding of its function. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service