Skip to Content
Merck
All Photos(1)

Key Documents

574711

Sigma-Aldrich

Syk Inhibitor

The Spleen Tyrosine Kinase Inhibitor, also referenced under CAS 622387-85-3, controls the biological activity of Spleen Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Syk Inhibitor, Spleen Tyrosine Kinase Inhibitor, 3-(1-Methyl-1H-indol-3-yl-methylene)-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O3S
CAS Number:
Molecular Weight:
353.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (sum of two isomers, HPLC)

form

solid

potency

14 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

deep orange

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1cc2c(cc1)NC(=O)C2=Cc3c4c([n](c3)C)cccc4

InChI

1S/C18H15N3O3S/c1-21-10-11(13-4-2-3-5-17(13)21)8-15-14-9-12(25(19,23)24)6-7-16(14)20-18(15)22/h2-10H,1H3,(H,20,22)(H2,19,23,24)

InChI key

MLKHXLFEYOOYEY-UHFFFAOYSA-N

General description

A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor (IC50 = 14 nM). Has been shown to effectively inhibit FcεRI-mediated degranulation in the rat basophilic leukemia cell line, RBL-2H3 (EC50 = 313 nM).
A cell-permeable, potent, reversible, and ATP-competitive Spleen Tyrosine Kinase (Syk) inhibitor (IC50 = 14 nM). Shown to effectively inhibit FcεRI-mediated degranulation in the rat basophilic leukemia cell line, RBL-2H3 (EC50 = 313 nM).

Biochem/physiol Actions

Cell permeable: yes
EC50 = 313 nM against FcεRI-mediated degranulation in the rat basophilic leukemia cell line, RBL-2H3
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Lai, J.Y.Q., et al. 2003. Bioorg. Med. Chem. Lett.13, 3111.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tai Yeon Koo et al.
Cell biology international, 37(1), 19-28 (2013-01-16)
Atherosclerosis develops from cascades of inflammatory processes. Spleen tyrosine kinase (Syk) and monocyte chemotatic protein-1 (MCP-1) play important roles in the pathogenesis of atherosclerosis. Mycophenolic acid (MPA) has an anti-inflammatory effect. We have investigated whether MPA regulates Syk to repress
Patricia Vuscan et al.
Frontiers in immunology, 15, 1323333-1323333 (2024-02-28)
Candida albicans cell wall component β-glucan has been extensively studied for its ability to induce epigenetic and functional reprogramming of innate immune cells, a process termed trained immunity. We show that a high-complexity blend of two individual β-glucans from Saccharomyces
Genome-Wide Association Study Reveals CLEC7A and PROM1 as Potential Regulators of Paracoccidioides brasiliensis-Induction of Cytokine Production in Peripheral Blood Mononuclear Cells.
de Figueiredo, et al.
Journal of Fungi, 9 (2023)
Lin Wang et al.
Nature genetics, 53(4), 500-510 (2021-03-31)
Spleen tyrosine kinase (SYK) is a critical immune signaling molecule and therapeutic target. We identified damaging monoallelic SYK variants in six patients with immune deficiency, multi-organ inflammatory disease such as colitis, arthritis and dermatitis, and diffuse large B cell lymphomas.
Omar Rafael Alemán et al.
Frontiers in immunology, 12, 657393-657393 (2021-06-01)
Human neutrophils express two unique antibody receptors for IgG, the FcγRIIa and the FcγRIIIb. FcγRIIa contains an immunoreceptor tyrosine-based activation motif (ITAM) sequence within its cytoplasmic tail, which is important for initiating signaling. In contrast, FcγRIIIb is a glycosylphosphatidylinositol (GPI)-linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service