Skip to Content
Merck
All Photos(1)

Key Documents

2100

Millipore

Protein-Concentrate Kit (Micro)

Protein-Concentrate Kit (Micro) is suitable for concentrating proteins for running gels, raising antibodies, protein purification, protein assays & other applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

manufacturer/tradename

Chemicon®

technique(s)

protein purification: suitable (concentration)

application(s)

sample preparation

shipped in

ambient

General description

The Chemicon Protein-Concentrate kit uses a proprietary reagent Universal Protein Precipitation Agent (UPPA). Protein solutions as dilute as 1 ng/mL can be quantitatively concentrated into a small volume. Protein precipitation is not effected by the presence of detergents, chaotropic, or other common laboratory agents. After precipitation, the precipitate is suspended in a small volume of Precipitate Suspension Buffer supplied with each kit. If the protocol is followed correctly, the recovery is generally 100%. There are enough reagents for concentrating up to 10 mL of dilute protein solution.

Packaging

10 samples

Components

UPPA-I 30 Ml

UPPA-II 30 mL

OrgoSol Buffer 50 mL

UPC-Wash 2.0 mL

UPC-SEED 0.3 mL

Precipitate Buffer I 2.0 mL

Precipitate Buffer II 0.5 mL

Storage and Stability

Store at room temperature for up to 6 months from date of purchase. To avoid possible microbial contamination, dispense all solutions aseptically.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Custom-designed MLPA using multiple short synthetic probes: application to methylation analysis of five promoter CpG islands in tumor and urine specimens from patients with bladder cancer.
Serizawa RR, Ralfkiaer U, Dahl C, Lam GW, Hansen AB, Steven K, Horn T, Guldberg P
The Journal of Molecular Diagnostics : JMD null
Ankita Anusruti et al.
Obesity facts, 13(1), 66-76 (2020-01-28)
In the literature, obesity is discussed as a determinant of high oxidative stress (OS). Hence, prevention or reduction of obesity could prevent high OS and subsequently serve as a target for "healthy aging." Diacron's reactive oxygen metabolites test (D-ROM) and
Eric W Refsland et al.
Nucleic acids research, 38(13), 4274-4284 (2010-03-24)
The human APOBEC3 proteins are DNA cytidine deaminases that impede the replication of many different transposons and viruses. The genes that encode APOBEC3A, APOBEC3B, APOBEC3C, APOBEC3D, APOBEC3F, APOBEC3G and APOBEC3H were generated through relatively recent recombination events. The resulting high
Tbr1 regulates regional and laminar identity of postmitotic neurons in developing neocortex.
Bedogni F, Hodge RD, Elsen GE, Nelson BR, Daza RA, Beyer RP, Bammler TK, Rubenstein JL, Hevner RF
Proceedings of the National Academy of Sciences of the USA null
Dendritic cell nuclear protein-1, a novel depression-related protein, upregulates corticotropin-releasing hormone expression.
Zhou T, Wang S, Ren H, Qi XR, Luchetti S, Kamphuis W, Zhou JN, Wang G, Swaab DF
Brain null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service