Skip to Content
Merck
All Photos(1)

Key Documents

208910

Sigma-Aldrich

Anti-Calreticulin (405-417) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

monkey, human, mouse, canine, hamster, bovine, rat, porcine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Calreticulin (405-417), rabbit polyclonal, recognizes the ~63 kDa calreticulin in heat-shocked HeLa cells. It is validated for WB, ICC, IHC, and IP.
Protein A purified rabbit polyclonal antibody. Recognizes the ~63 kDa calreticulin protein.
Recognizes the ~63 kDa calreticulin protein in heat-shocked HeLa cells.

Immunogen

Human
a synthetic peptide [(C)EEEDVPGQAKDEL] corresponding to amino acids 405-417 of human calreticulin, conjugated to KLH

Application

Immunoblotting (1:500, mouse/rat; 1:1000, human, chemiluminescent)

Immunocytochemistry (see comments)

Immunoprecipitation (see comments)

Immunohistochemistry (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Does not recognize calreticulin in Drosophila cells. This antibody has been reported to work for immunocytochemistry, immunohistochemistry, and immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.
Janicki, S., et al. 1997. J. Cell Biol. 139, 485.
Wada, I., et al. 1997. EMBO J. 16, 5420.
Nauseef, W.F., et al. 1995. J. Biol. Chem.270, 4741.
Jethmalani, S.M., et al. 1994. J. Biol. Chem. 269, 23603.
Smith, M.J., and Koch, G.L.E. 1989. EMBO J.8, 3581.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine A Fawcett et al.
Diabetes, 57(9), 2527-2533 (2008-07-02)
Loss of lipin 1 activity causes lipodystrophy and insulin resistance in the fld mouse, and LPIN1 expression and common genetic variation were recently suggested to influence adiposity and insulin sensitivity in humans. We aimed to conduct a comprehensive association study
Matthew J Rardin et al.
The Journal of biological chemistry, 283(22), 15440-15450 (2008-04-04)
Although large-scale approaches have identified numerous mitochondrial phosphoproteins, little is known about the mitochondrial kinases and phosphatases that regulate these phosphoproteins. Here, we identify two members of the atypical dual specificity phosphatases (DSP), DSP18 and DSP21, that are localized in
Cláudia Rosa-Ferreira et al.
Developmental cell, 21(6), 1171-1178 (2011-12-17)
Lysosomes move bidirectionally on microtubules, and this motility can be stimulated by overexpression of the small GTPase Arl8. By using affinity chromatography, we find that Arl8-GTP binds to the soluble protein SKIP (SifA and kinesin-interacting protein, aka PLEKHM2). SKIP was
Safa Lucken-Ardjomande Häsler et al.
The Journal of cell biology, 219(5) (2020-04-29)
In addition to the classical pathway of secretion, some transmembrane proteins reach the plasma membrane through alternative routes. Several proteins transit through endosomes and are exported in a Rab8-, Rab10-, and/or Rab11-dependent manner. GRAFs are membrane-binding proteins associated with tubules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service