Skip to Content
Merck
All Photos(1)

Key Documents

208722

Sigma-Aldrich

Calpain Inhibitor III

The Calpain Inhibitor III, also referenced under CAS 88191-84-8, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Calpain Inhibitor III, Carbobenzoxy-valinyl-phenylalaninal, MDL 28170

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4
CAS Number:
Molecular Weight:
382.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

potency

8 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 20 mg/mL (Use only anhydrous DMSO.)

shipped in

ambient

storage temp.

2-8°C

SMILES string

N([C@@H](C(C)C)C(=O)N[C@@H](Cc2ccccc2)C=O)C(=O)OCc1ccccc1

InChI

1S/C22H26N2O4/c1-16(2)20(24-22(27)28-15-18-11-7-4-8-12-18)21(26)23-19(14-25)13-17-9-5-3-6-10-17/h3-12,14,16,19-20H,13,15H2,1-2H3,(H,23,26)(H,24,27)/t19-,20-/m0/s1

InChI key

NGBKFLTYGSREKK-PMACEKPBSA-N

General description

A potent, cell-permeable inhibitor of calpain I and II (Ki = 8 nM) that also reduces capsaicin-mediated cell death in cultured dorsal root ganglion. Reported to block A23187-induced suppression of neurite outgrowth in isolated hippocampal pyramidal neurons. Exhibits neuroprotective effects against glutamate-induced toxicity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
calpain 1, calpain 2
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Val-Phe-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Chatterjee, S., et al. 1998. J. Med. Chem.41, 2663.
Rami, A., et al. 1997. Neurosci. Res. 27, 93.
Yamazaki, T., et al. 1997. Biochemistry36, 8377.
Klafki, H., et al. 1996. J. Biol. Chem.271, 28655.
Chard, P.S., et al. 1995. Neuroscience 65, 1099.
Song, D.K., et al. 1994. J. Neurosci. Res. 39, 474.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vimala Bondada et al.
Biochimica et biophysica acta. Molecular cell research, 1868(7), 119019-119019 (2021-04-04)
The enzymatic characteristics of the ubiquitous calpain 5 (CAPN5) remain undescribed despite its high expression in the central nervous system and links to eye development and disease. CAPN5 contains the typical protease core domains but lacks the C terminal penta-EF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service