Skip to Content
Merck
All Photos(1)

Key Documents

203724

Millipore

Brij® 35 Detergent, 30% Aqueous Solution

Non-ionic detergent useful for the isolation of functional membrane complexes. Aggregation number: 40; HLB number: 16.9. Note: product liquifies upon warming to 50°C.

Synonym(s):

Polyoxyethyleneglycol Dodecyl Ether, Polyoxyethylene (23) Lauryl Ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

description

Merck USA index - 14, 7558

Quality Level

form

gel form

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

40

color

clear

CMC

92 μM

shipped in

ambient

storage temp.

15-25°C

SMILES string

O(CCO)CCCCCCCCCCCC

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

General description

Non-ionic detergent useful for the isolation of functional membrane complexes (aggregation number: 40; CMC: 92 mM; HLB number: 6.9). Product liquefies upon warming to 50°C.
Note: product liquifies upon warming to 50°C.

Application


  • Effect of microemulsions on transdermal delivery of citalopram: optimization studies using mixture design and response surface methodology.: This study explores the use of microemulsions, including Brij® 35, to enhance the transdermal delivery of citalopram. The researchers optimized the formulation using mixture design and response surface methodology, highlighting the effectiveness of Brij® 35 in improving drug delivery (Huang et al., 2013, Huang et al., 2013).

  • Activity and stability of catalase in nonionic micellar and reverse micellar systems.: This article examines the activity and stability of catalase in the presence of nonionic micellar and reverse micellar systems, including Brij® 35. The study demonstrates how Brij® 35 can stabilize enzyme activity, suggesting its potential application in biochemical assays and industrial processes (Gebicka & Jurgas-Grudzinska, 2004, Gebicka & Jurgas-Grudzinska, 2004).

Warning

Toxicity: Harmful (C)

Physical form

30% aqueous solution.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service