Skip to Content
Merck
All Photos(2)

Key Documents

1.71202

Supelco

Thermoreactor

number of holes 2X12 hole, Spectroquant® TR 620

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
SE.11

manufacturer/tradename

Spectroquant® TR 620

technique(s)

photometry: suitable

storage temp.

2-30°C

General description

2 x 12 holes in 2 heating zones, temperature and time separately controllable for each zone, 7 preinstalled programmes and 8 freely selectable programmes TR 620 Spectroquant®
The Spectroquant Thermoreactor TR 620 is designed for the digestion up to 24 samples simultaneously. Precisely controlled thermal digestion is a key sample preparation step during the determination of COD, TOC, total contents of cadmium, chromium, cyanide, iron, lead, nickel, nitrogen, phosphorus, silver, or zinc using our high quality Spectroquant Test Kits.

The Spectroquant Thermoreactor TR 620 features two separate heating blocks, which can be individually controlled. Thereby, the Thermoreactor TR 620 enables users to set up and perform two different digestion programs at the same time – enhancing flexibility and reducing waiting times.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service