Skip to Content
Merck
All Photos(1)

Key Documents

1.70374

Supelco

Chromium ICP standard

traceable to SRM from NIST Cr(NO₃)₃ in HNO₃ 2-3% 10000 mg/l Cr Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

product line

Certipur®

concentration

10000 mg/L Cr in 2-3% HNO3

technique(s)

ICP: suitable

pH

<1 (20 °C in H2O)

density

1.046 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Chromium ICP standard can be used as a calibration standard in the determination of chromium in contaminated water by inductively coupled plasma optical emission spectroscopy (ICP-OES).

Analysis Note

10000 mg/L Cr in 2-3% nitric acid, preparedwith high-purity Cr(NO3)3, HNO3, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined adsorption-permeation membrane process for the removal of chromium (III) ions from contaminated water
Pagana A, et al.
Journal of Membrane Science, 367, 319-324 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service