Skip to Content
Merck
All Photos(1)

Documents

1.70338

Supelco

Osmium ICP standard

(NH₄)₂OsCl₆ in HCl 7% 1000 mg/l Os Certipur®

Synonym(s):

Osmium Calibration Standard for ICP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

density

1.034 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

The osmium ICP standard may find the below uses:
  • To study the spatial distribution of isosteric osmium and ruthenium anticancer agents using laser ablation-inductively coupled plasma-mass spectrometry (LA-ICP-MS)
  • Determination of osmium by inductively coupled plasma-mass spectrometry (ICP-MS) after pressure vessel digestion with nitric acid
  • Development of a plasma induced vapor generation method to measure osmium in solutions by inductively coupled plasma-optical emission spectrometry (ICP-OES)
  • Study of serum-binding properties of two metallodrugs by size exclusion chromatography-inductively coupled plasma-mass spectrometry (SEC-ICP-MS)
  • Detection of osmium by Photochemical Vapor Generation-Inductively Coupled Plasma-Mass Spectrometry(PVG-ICP-MS)
  • Inductively coupled plasma-optical emission spectrometric (ICP-OES) determination of osmium by using photooxidation vapor generation (POVG) method

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum-binding properties of isosteric ruthenium and osmium anticancer agents elucidated by SEC-ICP-MS
Klose M, et al.
Monatshefte fur Chemie / Chemical Monthly, 149(10), 1719-1726 (2018)
Bioimaging of isosteric osmium and ruthenium anticancer agents by LA-ICP-MS
Klose M, et al.
Metallomics : Integrated Biometal Science, 10(3), 388-396 (2018)
On line vapor generation of osmium based on solution cathode glow discharge for the determination by ICP-OES
Zhu Z, et al.
Talanta, 106, 133-136 (2013)
High-efficiency photooxidation vapor generation of osmium for determination by inductively coupled plasma-optical emission spectrometry
Zhu Z, et al.
Journal of Analytical Atomic Spectrometry, 29(3), 506-511 (2014)
Simultaneous Detection of Ruthenium and Osmium by Photochemical Vapor Generation-Inductively Coupled Plasma-Mass Spectrometry
Yang Q, et al.
Analytical Chemistry (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service