Skip to Content
Merck
All Photos(1)

Key Documents

16-321

Sigma-Aldrich

Anti-KRAS Antibody

mouse monoclonal

Synonym(s):

GTP- and GDP-binding peptide B, GTPase Hras, Ha-Ras1 proto-oncoprotein, Ras family small GTP binding protein H-Ras, Transforming protein p2, c-has/bas p21 protein, c-ras-Ki-2 activated oncogene, p19 H-RasIDX protein, transformation gene: oncogene HAMSV,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Ras Magnetic Bead Conjugate Antibody, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The tumor oncoproteins HRas, KRas, and NRas are rated as Ras protoncogenes. Point mutations within Ras genes are frequently detected in human malignancies and in experimentally induced tumors in animals.

Specificity

Other species unknown.
This antibody recognizes p21H-, K- and N-Ras.

Immunogen

KLH-conjugated protein corresponding to human Ras.

Application

Anti-Ras Magnetic Bead Conjugate Antibody detects level of Ras Magnetic Bead Conjugate & has been published & validated for use in WB & IP.
Immunoprecipitation:
150 μg of this antibody has been reported to immunoprecipitate Ras.
Research Category
Signaling
Research Sub Category
MAP Kinases

G-proteins

Target description

21 kDa

Physical form

20% slurry; 1xPBS, 0.05% sodium azide. Sufficient for 10 immunoprecipitation assays.
Format: Purified
Protein G Purified

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
DO NOT FREEZE

Analysis Note

Control
A431 whole cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service