Skip to Content
Merck
All Photos(1)

Documents

1.46762

Millipore

ReadyPlate 55 KIT TGE Agar

suitable for filtration, microbiology, box of 150 sets (55 mm plate and EZ-PAK membrane filter)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

gridded white mixed cellulose esters (MCE) membrane

Quality Level

Agency

ISO

form

plate (ready-to-use)

feature

ready-to-use

packaging

box of 150 sets (55 mm plate and EZ-PAK membrane filter)

manufacturer/tradename

ReadyPlate

technique(s)

cell counting: suitable (total viable count (TVC))

pore size

0.45 μm pore size

application(s)

filtration
microbiology

food and beverages

storage temp.

15-25°C

suitability

nonselective for

General description

Application: Enumeration of total viable count, Membrane filtration method

Packaging: 55 mm plate and EZ PAK membrane filter (150 sets per box)

Synonym: Tryptone Glucose Extract Agar, Tryptone Glucose Meat Extract Agar

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service