Skip to Content
Merck
All Photos(1)

Key Documents

1.44111

Millipore

Cover Slides

HYCON®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113035
NACRES:
NA.74
Pricing and availability is not currently available.

form

plate (ready-to-use)

Quality Level

feature

ready-to-use

packaging

pkg of 100 units

manufacturer/tradename

HYCON®

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with Agar Strips

storage temp.

15-25°C

General description

Cover slides are used with the non-irradiated agar strips, for the detection of airborne microorganisms with RCS® Air Samplers. The cover slides are used to cover the agar surface post sampling and during incubation to avoid cross contamination and false positive results.

Application

Cover slides covers the agar strip surface and prevents contamination post sampling and during incubation.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service