Skip to Content
Merck
All Photos(1)

Key Documents

1.19754

Supelco

di-Potassium hydrogen phosphate trihydrate

buffer substance for chromatography LiChropur

Synonym(s):

di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HK2O4P · 3H2O
CAS Number:
Molecular Weight:
228.22
MDL number:
UNSPSC Code:
12161703
EC Index Number:
231-834-5
NACRES:
NA.21
grade:
reagent grade
form:
solid
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

technique(s)

HPLC: suitable

pH

9.2-9.4 (20 °C, 50 g/L in H2O)

mp

340 °C (anhydrous substance; decomposes)

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[K+].[P](=O)([O-])([O-])O.O.O.O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

Analysis Note

Assay (acidimetric): ≥ 99.0 %
pH-value (5 %; water): 9.2 - 9.4
Self-absorption (254 nm; 0.2 mol/l; 1 cm; water): ≤ 0.01
Self-absorption (220 nm; 0.2 mol/l; 1 cm; water): ≤ 0.03
Self-absorption (215 nm; 0.2 mol/l; 1 cm; water): ≤ 0.11

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service