Skip to Content
Merck
All Photos(1)

Key Documents

1.14848

Supelco

Phosphate Test, photometric

photometric, 0.0025-5.00 mg/L (PO4-P), 0.0077-15.3 mg/L (PO43-), 0.0057-11.46 mg/L (P2O5), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Phosphate Test, photometric, 0.0025-5.00 mg/L (PO4-P), 0.0077-15.3 mg/L (PO43-), 0.0057-11.46 mg/L (P2O5), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 220 tests (depending on package size)
sufficient for 420 tests

specific analyte(s)

phosphate

measuring range

0.0025-5.00 mg/L (PO4-P)
0.0057-11.46 mg/L (P2O5)
0.0077-15.3 mg/L (PO43-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phosphomolybdenum blue)

storage temp.

15-25°C

General description

(o-phosphate) Method: photometric, PMB 0.0025 - 5.00 mg/l PO₄-P 0.0077 - 15.3 mg/l PO₄ 0.0057 - 11.46 mg/l P₂O₅ Spectroquant®
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant®Phosphate Reagent Test allows the accurate quantification of phosphate. Elevated levels of phosphate might lead to eutrophication of water bodies, which is a risk to the environment.
Therefore, the concentration of phosphate in drinking water and wastewater is regulated in many countries. Further applications are the determination of phosphate in foods & beverages (e.g. beer) and in environmental and agricultural samples (e.g. soil).
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi, Do you have any data regarding the solubility of the p2K reagent (mix of Molybdate and antimony tartrate)?

    1 answer
    1. The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/198/700/114848ug-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service