Skip to Content
Merck
All Photos(1)

Documents

1.14547

Supelco

Nitrite Test, photometric

photometric, 0.03-2.30 mg/L (NO2-), 0.010-0.700 mg/L (NO2-N), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrite Cell Test, photometric, 0.03-2.30 mg/L (NO2-), 0.010-0.700 mg/L (NO2-N), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

measuring range

0.010-0.700 mg/L (NO2-N)
0.03-2.30 mg/L (NO2-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Griess’ reaction)

storage temp.

15-25°C

General description

Method: photometric 0.03 - 2.30 mg/l NO₂⁻ 0.010-0.700 mg/l NO₂-N Spectroquant®
The method is analogous to EPA 354.1, APHA 4500-NO2 - B, and DIN EN 26777.
This Spectroquant® Nitrite Cell Test allows the accurate quantification of the nitrite content in aqueous samples.
Method applied: In acidic solution nitrite ions react with sulfanilic acid to form a diazonium salt. This reacts with N-(1-naphthyl)ethylenediamine dihydrochloride to form a red-violet azo dye that is determined photometrically.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided .
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2DD barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Application


  • Bronchom assuages airway hyperresponsiveness in house dust mite-induced mouse model of allergic asthma and moderates goblet cell metaplasia, sub-epithelial fibrosis along with changes in Th2 cytokines and chemokines.: This study discusses the nitrite cell test in evaluating goblet cell metaplasia and sub-epithelial fibrosis in a mouse model of asthma, providing insights into cellular changes and cytokine responses (Balkrishna et al., 2024).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service