Skip to Content
Merck
All Photos(4)

Key Documents

1.10337

Supelco

Peroxide Test Strips, colorimetric

greener alternative

100-1000 mg/L (H₂O₂), MQuant®

Synonym(s):

Peroxide chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.21

Product Name

Peroxide Test, colorimetric, 100-1000 mg/L (H2O2), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: enzymatic reaction
reagent type: oxidant

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

100-1000 mg/L (H2O2)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: colorimetric with test strips 100 - 200 - 400 - 600 - 800 - 1000 mg/l (H2O2) MQuant®

Peroxide is an all purpose disinfectant which, due to its excellent antiviral, bactericidal and fungicidal action, is widely used in the food industry. Peroxidase transfers peroxide oxygen to an organic redox indicator. This produces a yellow-brown oxidation product. The peroxide concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. Each strip also features a second reaction zone (alert zone). This zone changes color in the presence of high peroxide concentrations (from approx. 2000 mg/l (H2O2) on) and of interfering concentrations of peracetic acid. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peroxide concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Peroxide test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Food Quality Control: In the food industry, peroxide tests are used to determine the peroxide value in vegetable oils and other edible fats. This application is crucial for assessing the freshness and shelf life of food products. A recent study demonstrated the use of paper-based analytical devices for the colorimetric determination of peroxide values in vegetable oils, highlighting the simplicity and effectiveness of this method for routine food quality testing (Ghohestani et al., Food Chemistry, 2023).

Linkage

Replaces: 10337-1; 10337

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service