1.09921
Potassium hydroxide solution
c(KOH) = 0.1 mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®
About This Item
Recommended Products
product name
Potassium hydroxide solution, for 1000 ml, c(KOH) = 0.1 mol/l (0.1 N) Titrisol®
Quality Level
product line
Titrisol®
form
liquid
reaction suitability
reaction type: Acid-base reactions
concentration
0.1 M
technique(s)
titration: suitable
pH
14 (20 °C in H2O)
density
1.09 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
[K+].[O-][H]
InChI
1S/K.H2O/h;1H2/q+1;/p-1
InChI key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
- Voltammetric Determination: Potassium hydroxide solution in ethanol is used in voltammetric determination of antioxidants in various matrices. A recent study demonstrated its application in the voltammetric determination of the antioxidant 2,6-di-tert-butyl-p-cresol (T501) in transformer oil using a graphite electrode. This method enhances the sensitivity and accuracy of antioxidant detection, crucial for ensuring the stability and performance of oils and other industrial fluids (Zhou et al., Analytical Letters, 2021).
- Oxidative Degradation Studies: The reagent is employed in oxidative degradation studies of organic compounds. For instance, it has been used in the oxidative degradation of piperazine in potassium hydroxide solution for CO2 absorption processes. This application is essential for developing efficient CO2 capture and storage technologies, contributing to environmental sustainability and climate change mitigation (Ochedi et al., SSRN Electronic Journal, 2022).
- CO2 Absorption and Modeling: Potassium hydroxide solution in ethanol is used in experimental and modeling studies of CO2 absorption processes. Recent research has focused on the absorption of CO2 into potassium hydroxide aqueous solution, providing valuable data for optimizing industrial CO2 capture methods. This application is crucial for enhancing the efficiency and effectiveness of carbon capture technologies (Rastegar and Ghaemi, Heat and Mass Transfer, 2022).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service