1.09533
pH-indicator strips pH 5.0 - 10.0
non-bleeding, colorimetric, pH range 5.0-10.0, graduations and accuracy accuracy: 0.5 pH unit, MQuant®
Synonym(s):
pH stick pH 5.0 - 10.0, pH test strips pH 5.0 - 10.0
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41113034
NACRES:
NB.35
Recommended Products
Agency
suitable for SM 5210
Quality Level
product line
MQuant®
feature
non-bleeding
graduations and accuracy measuring range
accuracy: 0.5 pH unit
pH range
5.0-10.0
detection method
colorimetric
storage temp.
2-30°C
General description
The MQuant pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the three reaction zones on the test strip with the fields of the color scale, perfectly matching the reaction color. The clever Safetyedge boxes allow for safe storage and convenient withdrawal of the test strips. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant, see instructions of use), we recommend the use of MQuant pH test strips as pre-test.
non-bleeding pH 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 - 9.5 - 10 0 MQuant®
Linkage
Replaces: 9588-1; 9588-3; 9588
Analysis Note
Application test: conforms
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service