Skip to Content
Merck
All Photos(1)

Key Documents

1.07861

Sigma-Aldrich

Strontium carbonate

Technipur®

Synonym(s):

Carbonic acid strontium salt, Strontianite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

pH

7-8 (20 °C, 0.01 g/L in H2O)

mp

1497 °C

density

3.7 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium carbonate is a precursor material used in the manufacture of colorants in fireworks, special glasses, computer monitors, paints, glass cathode-ray tubes, medicine, and photocatalysts.

Application

Strontium carbonate can be used to prepare:
  • (1-x)(Bi0.5Na0.5)TiO3-xSrTiO3-ceramics exhibiting piezoelectric properties.
  • Samarium and sodium-doped SrBi4Ti4O15 piezoelectric ceramics.
  • Sodium and praseodymium substituted Sr0.2Na0.4Pr0.4Bi4Ti4O15 piezoelectric ceramic for high-temperature sensor applications.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dielectric properties of Na and Pr doped SrBi4Ti4O15 ceramics
Rajashekhar G
Materials Today, 33(8), 5467-5470 (2020)
Effect of Sm and Na substitution on dielectric properties of SrBi4Ti4O15
Ravikiran U
Ferroelectrics, 537(1) (2018)
Piezoelectric properties and phase transition temperatures of the solid solution of (1 - x)(Bi0.5Na0.5)TiO3?xSrTiO3
Krauss W
J. Eur. Ceram. Soc., 30(8), 1827-1832 (2010)
Visible Light Responsive Strontium Carbonate Catalyst Derived from Solvothermal Synthesis
Wichannananon P, et al.
Catalysts, 10(9), 1069-1069 (2020)
Preparation of nano-structured strontium carbonate from Dasht-e kavir celestite ore via mechanochemical method
Alimohammadi E, et al.
Journal of Ultrafine Grained and Nanostructured Materials, 51(2), 147-152 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service