1.02449
Readybag® Half FRASER broth with supplements
pkg of 60 pieces, according to FDA (BAM), ISO 11290, for Listeria spp.
Synonym(s):
Demi FRASER broth with supplements
About This Item
Agency
APHA
ISO 11290
Quality Level
reg. compliance
according to FDA (BAM)
sterility
sterile; γ-irradiated
form
medium granules (dehydrated (DCM))
packaging
pkg of 60 pieces
pkg of
manufacturer/tradename
ReadyBag®
technique(s)
microbiological culture: suitable
pH
7.0-7.4 (25 °C, 55 g/L in H2O)
solubility
55 g/L
application(s)
food and beverages
microbiology
pathogen testing
storage temp.
15-25°C
suitability
Listeria spp. (spp.)
General description
The high nutrient content and the large buffer capacity of the Readybag® Half FRASER broth (with supplements) create optimum growth conditions for Listeria spp. Lithium chloride, nalidixic acid, and acriflavine hydrochloride inhibit the growth of accompanying bacteria. Esculin and ammonium iron (III) citrate forms a black complex of esculetin with iron (III) ions and facilitates the detection of β-D-glucosidase activity. But this reaction is not exclusive to Listeria spp. Hence, every primary and secondary enrichment in Fraser broth must be sub-cultured on selective plating media.
Application
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service