Skip to Content
Merck
All Photos(2)

Key Documents

1.01514

Supelco

Hydrochloric acid

30%, Ultrapur

Synonym(s):

Hydrochloric acid solution, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Hydrochloric acid solution, 30% Ultrapur

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

vapor pressure

21.8 hPa ( 20 °C)

Assay

30%

form

liquid

pH

<1 (20 °C, 300 g/L in H2O)

bp

85 °C/1013 hPa

mp

-50 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Corrosion inhibition of mild steel in hydrochloric acid solution by the expired Ampicillin drug.: This study investigates the use of expired Ampicillin as a corrosion inhibitor for mild steel in hydrochloric acid solutions, showcasing an innovative approach to repurposing pharmaceutical waste (Alamry et al., 2023).

  • Determination of Carcinogenic Primary Aromatic Amines Contained as Impurities in Synthetic Organic Colorants.: This research presents a method for detecting carcinogenic primary aromatic amines in synthetic organic colorants using hydrochloric acid solutions for sample preparation, contributing to enhanced safety in consumer products (Kawakami et al., 2022).

  • Determination of Amino Acid Composition of Ganirelix Acetate in an Injectable Formulation by Pre-column Derivatization with 6-Aminoquinolyl-N-hydroxysuccinimidyl Carbamate.: This study describes the determination of amino acids in Ganirelix Acetate formulations using hydrochloric acid solutions, providing a detailed analytical method for pharmaceutical applications (Ummiti and Kumar, 2020).

  • In-process prepared deep eutectic solvent based homogeneous liquid-liquid microextraction for the determination of irgaphos 168 and irganox 1010 in polypropylene packed drinks.: The research focuses on using hydrochloric acid solutions in the extraction process for determining antioxidants in drinks, enhancing food safety and quality control methods (Naebi et al., 2020).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service