Skip to Content
Merck
All Photos(1)

Key Documents

1.01056

Supelco

Aluminium

fine powder, stabilized about 2% fat

Synonym(s):

Aluminium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12141702
EC Index Number:
231-072-3
NACRES:
NA.21

Quality Level

Assay

≥90.0% (complexometric)

autoignition temp.

400 °C

impurities

≤0.0005% As (Arsenic)
≤0.03% Heavy metals (as Pb)
≤1.0% Fe (Iron)

bp

2467 °C/1013 hPa

mp

660 °C

density

2.70 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Related Categories

Application


  • A recyclable luminescent MOF sensor for on-site detection of insecticide dinotefuran and anti-Parkinson′s drug entacapone in various environmental and biological specimens.: This study highlights the development of a metal-organic framework (MOF) sensor that can detect traces of dinotefuran and entacapone, demonstrating potential applications in environmental monitoring and healthcare diagnostics (Mir NUD et al., 2024).

  • A lithium-aluminium heterobimetallic dimetallocene.: This paper introduces a novel bimetallic compound combining lithium and aluminium, offering insights into their cooperative effects in chemical synthesis and potential applications in material science (Bischoff IA et al., 2024).

Analysis Note

Assay (complexometric): ≥ 90.0 %
Heavy metals (as Pb): ≤ 0.03 %
As (Arsenic): ≤ 0.0005 %
Fe (Iron): ≤ 1.0 %
Fat: passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service