1.00923
Diethyl ether
≥99% (GC), EMPLURA®
Synonym(s):
Et2O, Ethoxyethane, Ethyl ether, Ether, Ether, Ethyl ether
About This Item
Recommended Products
product name
Diethyl ether, EMPLURA®
vapor density
2.6 (vs air)
Quality Level
vapor pressure
587 hPa ( 20 °C)
product line
EMPLURA®
Assay
≥99% (GC)
form
liquid
autoignition temp.
320 °F
potency
1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
expl. lim.
36.5 %
impurities
≤0.00003% Peroxide (as H2O2)
≤0.002% Free acid (as CH3COOH)
≤0.2% Water
refractive index
n20/D 1.3530 (lit.)
bp
34.6 °C (lit.)
mp
−116 °C (lit.)
transition temp
flash point -40 °C
density
0.706 g/mL at 25 °C (lit.)
storage temp.
15-25°C
SMILES string
CCOCC
InChI
1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3
InChI key
RTZKZFJDLAIYFH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Determination of chloropropanol esters and glycidyl esters in nutritional foods by gas chromatography-tandem mass spectrometry based on acid hydrolysis and solid-phase extraction.: This study presents an innovative method for detecting toxic contaminants in food, utilizing gas chromatography with diethyl ether as a key solvent in sample preparation (Li et al., 2024).
- Cloning, Expression, Characterization and Immobilization of a Recombinant Carboxylesterase from the Halophilic Archaeon, Halobacterium salinarum NCR-1.: This research discusses the use of diethyl ether in the purification process of enzymes from extreme environments, enhancing biotechnological applications (Ortega-de la Rosa et al., 2024).
- Chemical composition and antimicrobial study of Crossobamon orientalis body oil.: Highlights the role of diethyl ether in extracting and analyzing the chemical constituents of biological samples, providing insights into antimicrobial properties (Ahmad et al., 2024).
- A Static Headspace Gas Chromatography Method for Quantitation of Residual Solvents in Nanoformulations.: Introduces a methodological advancement in analytical chemistry using diethyl ether for the quantification of residual solvents in pharmaceutical preparations, ensuring safety and compliance (Kattel and Clogston, 2024).
Packaging
Analysis Note
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.002 %
Density (d 20 °C/ 4 °C): 0.713 - 0.716
Peroxide (as H₂O₂): ≤ 0.00003 %
Readily carbonizable substances: conforms
Water: ≤ 0.2 %
Stabilized with 5 - 10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT)
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-40.0 °F - closed cup
Flash Point(C)
-40 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service