Skip to Content
Merck
All Photos(1)

Key Documents

07-687

Sigma-Aldrich

Anti-Cyclin E Antibody

Upstate®, from rabbit

Synonym(s):

Anti-CCNE, Anti-pCCNE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCNE1(898)

General description

Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are only expressed in tumors but not in normal tissue, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in the Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.

Specificity

Cyclin E

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 378-396 (GVLTPPHSSKKQSSEQETE) of rat Cyclin E

Application

Detect Cyclin E with Anti-Cyclin E Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IP, WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on RIPA lysates from 3T3/A31 cells

Target description

Mr 52kDa

Linkage

Replaces: 04-223

Physical form

ImmunoAffinity Purified
PBS containing 0.1% sodium azide and 0.2% gelatin.

Storage and Stability

stable for 1 year at 4°C from date of shipment

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of cyclin E in the regulation of entry into S phase.
Sauer, K and Lehner, C F
Progress in Cell Cycle Research, 1, 125-139 (1995)
Philipp Schuler et al.
Plant, cell & environment, 45(1), 12-22 (2021-09-27)
The analysis of the non-exchangeable hydrogen isotope ratio (δ2 Hne ) in carbohydrates is mostly limited to the structural component cellulose, while simple high-throughput methods for δ2 Hne values of non-structural carbohydrates (NSC) such as sugar and starch do not
Estrogen-induced activation of Cdk4 and Cdk2 during G1-S phase progression is accompanied by increased cyclin D1 expression and decreased cyclin-dependent kinase inhibitor association with cyclin E-Cdk2.
Prall, O W, et al.
The Journal of Biological Chemistry, 272, 10882-10894 (1997)
Cyclin A: function and expression during cell proliferation.
Desdouets, C, et al.
Progress in Cell Cycle Research, 1, 115-123 (1995)
Cell cycle regulation in mouse heart during embryonic and postnatal stages.
Aiko Ikenishi,Hitomi Okayama,Noriko Iwamoto,Satoshi Yoshitome,Shoji Tane,Kazuomi Nakamura et al.
Development, Growth & Differentiation null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service