Skip to Content
Merck
All Photos(3)

Key Documents

07-450

Sigma-Aldrich

Anti-monomethyl-Histone H3 (Lys9) Antibody

Upstate®, from rabbit

Synonym(s):

H3K9me1, Histone H3 (mono methyl K9), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

chicken, mouse, human

manufacturer/tradename

Upstate®

technique(s)

dot blot: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys9)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species cross-reactivity is expected based on sequence homology.
Recognizes monomethylated histone H3 (Lys9), Mr ~17 kDa.

Immunogen

Epitope: Lys9
KLH-conjugated, synthetic 2X-branched peptide containing the sequence …ARmeKST… in which meK corresponds to monomethyl-lysine at residue 9 of human histone H3.

Application

Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in ICC, PIA & WB.
Peptide Competition:
Specificity confirmed by the ability of 10 μM of the immunizing peptide to abolish detection of histone H3 in immunoblot analysis of HeLa acid extracts (Figure B, Lane 2). No signal reductions were detected with preincubation with 10 μM di- or trimethyl-lysine 9 or di- or trimethyl-lysine 27 modified peptides. Some signal reduction detected after preincubation with 10 μM monomethyl-lysine 27 modified peptide (Figure B, Lane 5, page 2).

Immunocytochemistry & Dot Blot: Reported by an independent laboratory1-4.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by Western Blot on acid extracted proteins from HeLa cells, but did not detect unmethylated recombinant Histone H3.

Western Blot Analysis:
0.1-0.2 μg/mL had detected monomethylated histone H3 in acid extracted proteins from HeLa cells, but did not detect unmethylated recombinant Histone H3 (Catalog # 14-411).

Target description

17 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit IgG in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominica Willmann et al.
International journal of cancer, 131(11), 2704-2709 (2012-03-27)
Post-translational modifications of histones by chromatin modifying enzymes regulate chromatin structure and gene expression. As deregulation of histone modifications contributes to cancer progression, inhibition of chromatin modifying enzymes such as histone demethylases is an attractive therapeutic strategy to impair cancer
Gabi Gerlitz et al.
Traffic (Copenhagen, Denmark), 8(11), 1521-1529 (2007-09-08)
Directed cell migration is a property central to multiple basic biological processes. Here, we show that directed cell migration is associated with global changes in the chromatin fiber. Polarized posttranslational changes in histone H1 along with a transient decrease in
Laura Pufahl et al.
Journal of cellular and molecular medicine, 16(7), 1461-1473 (2011-09-03)
The 5-lipoxygenase (5-LO) is the key enzyme in the formation of leukotrienes. We have previously shown that the histone deacetylase (HDAC) inhibitor trichostatin A (TSA) activates 5-LO transcription via recruitment of Sp1, Sp3 and RNA polymerase II to the proximal
Makoto Tachibana et al.
The EMBO journal, 27(20), 2681-2690 (2008-09-27)
Methylation of DNA and lysine 9 of histone H3 (H3K9) are well-conserved epigenetic marks for transcriptional silencing. Although H3K9 methylation directs DNA methylation in filamentous fungi and plants, this pathway has not been corroborated in mammals. G9a and GLP/Eu-HMTase1 are
Direct interaction between DNMT1 and G9a coordinates DNA and histone methylation during replication.
Esteve, PO; Chin, HG; Smallwood, A; Feehery, GR; Gangisetty, O; Karpf, AR; Carey, MF; Pradhan, S
Genes & Development null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service