Skip to Content
Merck
All Photos(4)

Key Documents

07-1417

Sigma-Aldrich

Anti-SGLT-1 (Sodium Glucose Co-transporter-1) Antibody

serum, from rabbit

Synonym(s):

Sodium/glucose cotransporter 1, Solute carrier family 5 member 1, Solute carrier family 5 (sodium/glucose cotransporter), member 1, Solute carrier family 5 (sodium/glucose transporter),member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

pig, rat, avian, rabbit, mouse, canine, human, reptile, feline

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC5A1(6523)

General description

The kidneys play a major role in the regulation of blood glucose. Although kidney nephrons indiscriminatly filter about 180g of glucose per day from the blood, most is reabsorbed actively in the proximal convoluted tubule by a Na+/Gl co-transport (ratio 2:1) mechanism (SGLT). Typically, glucose is first reabsorbed by these transporters, which reside within the brush-border epithelia of the PCT cells. The glucose is then transported out of the cell across the basolateral membranes by a facilitated diffusion sugar transporter (GLUTs). At least three members of SGLT′s (SGLT1-3) have been cloned and characterized from various species. SGLT1-3 genes code for protein of approximately 659-672 residues, with both N and C termini predicted to be extracellular. There is approximately 60-70% homology between the 3 forms. SGLT1 does not discriminate a-MDG, glucose and galactose, while SGLT2 and 3 do not transport D-galactose efficiently. SGLT1 is an integral membrane protein expressed in proximal convoluted tubule Sq1 segments of the kidney (outer renal medulla) and in the intestine.

Specificity

Recognizes SGLT-1.

Immunogen

Synthetic peptide corresponding to amino acids 402-420 in the cytoplasmic region of SGLT-1 of mouse/rabbit small intestine.

Application

Anti-SGLT-1 (Sodium Glucose Co-transporter-1) Antibody detects level of SGLT-1 (Sodium Glucose Co-transporter-1) & has been published & validated for use in WB, IH(P), ELISA.
Immunohistochemisty (paraffin): Anti-SGLT-1 staining on human kidney, tissue pretreated with citrate buffer, pH 6.0. A previous lot of this antibody was used at 1 μg/mL, using IHC-Select detection with HRP-DAB. Immunoreactivity is detected in the brush boarder of proximal convoluted tubule.

ELISA: 1:10,000 - 1:50,000 dilution of a previous lot, using 50-100 ng of control peptide/well.

Optimal working dilutions must be determined by end user.

Western Blot: A previous lot of this antibody was used on multiple lysates. See datasheet.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

Routinely evaluated by Western Blot on PC3 lysates.

Western Blot Analysis:
1:500-1:2,000 dilution of this lot detected SGLT-1 on 10 μg of PC3 lysates.

Target description

Recognizes a band of approximately 70-77 kDa in Western blot.

Linkage

Replaces: AB1352

Physical form

Rabbit polyclonal IgG antiserum in buffer containing 0.1% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt in undiluted aliquots.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
PC3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Verdile et al.
International journal of molecular sciences, 21(23) (2020-12-06)
We previously showed that, according to the frequency and distribution of specific cell types, the rainbow trout (RT) intestinal mucosa can be divided in two regions that form a complex nonlinear three-dimensional (3D) pattern and have a different renewal rate.
Glycosylation of the rabbit intestinal brush border Na+/glucose cotransporter.
Hirayama, B A and Wright, E M
Biochimica et Biophysica Acta, 1103, 37-44 (1992)
Tales Lyra Oliveira et al.
Scientific reports, 6, 21752-21752 (2016-02-24)
High glucose concentration in the airway surface liquid (ASL) is an important feature of diabetes that predisposes to respiratory infections. We investigated the role of alveolar epithelial SGLT1 activity on ASL glucose concentration and bacterial proliferation. Non-diabetic and diabetic rats
Saki Kondo et al.
Physiological reports, 7(20), e14255-e14255 (2019-10-28)
Long-term endurance training for a relatively short duration (~1 h) is reported to increase pancreatic amylase activity in rats, suggesting that chronic exercise training enhances carbohydrate digestive capacity. However, it remains unknown whether longer exercise training duration results in greater adaptation
Xuemin He et al.
Signal transduction and targeted therapy, 5(1), 45-45 (2020-04-30)
Previous studies by us and others demonstrated that activation of Wnt/β-catenin signaling plays a pathogenic role in chronic kidney diseases (CKD). Wnt co-receptor LRP5 variants are reported to associate with autosomal dominant polycystic kidney disease; but their exact roles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service