Skip to Content
Merck
All Photos(4)

Key Documents

07-1016

Sigma-Aldrich

Anti-phospho-Upf1 (Ser1127) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Regulator of nonsense transcripts 1, ATP-dependent helicase RENT1, Nonsense mRNA reducing factor 1, NORF1, Up-frameshift suppressor 1 homolog, hUpf1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), zebrafish (based on 100% sequence homology), yeast (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer1127)

Gene Information

human ... UPF1(5976)

General description

Regulator of nonsense transcripts 1 (UniProt: Q92900; also known as ATP-dependent helicase RENT1, Nonsense mRNA reducing factor 1, NORF1, Up-frameshift suppressor 1 homolog, hUpf1) is encoded by the UPF1 (also known as KIAA0221, RENT1) gene (Gene ID: 5976) in human. Upf1 is a member of the DNA2/NAM7 helicase family. It is a RNA-dependent helicase and ATPase required for nonsense-mediated decay (NMD) of mRNAs containing premature stop codons. It is recruited to mRNAs upon translation termination and undergoes a cycle of phosphorylation and dephosphorylation. It is phosphorylated by SMG1 and phosphorylation is considered as an essential step in NMD and is required for formation of mRNA surveillance complexes. Phosphorylated Upf1 is recognized by EST1B/SMG5, SMG6, and SMG7, which provide a link to the mRNA degradation machinery involving exonucleolytic and endonucleolytic pathways. A hyper-phosphorylated form is targeted to the P-body, while unphosphorylated protein is distributed throughout the cytoplasm. The ATPase activity of Upf1 is required for disassembly of mRNPs undergoing NMD and is also essential for embryonic viability. Two isoforms of Upf1 have been reported that are produced by alternative splicing. Upf1 contains a UPF1-type zinc-finger domain (aa 121-272) and a nucleotide-binding domain (aa 506-510).

Specificity

This rabbit polyclonal antibody detects Regulator of nonsense transcripts 1 (RENT1) in multiple species. It targets an epitope within 9 amino acids surrounding phosphorylated Ser1127.

Immunogen

Epitope: Phosphorylated Ser1127
KLH-conjugated linear peptide corresponding to Upf1 phosphorylated at Ser1127.

Application

Anti-phospho-Upf1 (Ser1127), Cat. No. 07-1016, is a highly specific rabbit polyclonal antibody that targets Regulator of nonsense transcripts 1 (RENT1) phosphorylated at Ser1127 and has been tested in Immunoprecipitation, peptide Inhibition Assay, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-Upf1 (Ser1127) in 10 µg of A431 treated with Calyculin A and Okadaic Acid.

Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot blocked phospho-Upf1 (Ser1127) in NIH3T3 treated with Calyculin A and Okadaic Acid.

Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated phospho-Upf1 (Ser1127) in NIH3T3 treated with Calyculin A and Okadaic Acid.

Quality

Evaluated by Western Blotting in NIH3T3 treated with Calyculin A and Okadaic Acid.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-Upf1 (Ser1127) in 10 µg lysate from NIH3T3 treated with Calyculin A (50 nM) and Okadaic Acid (500 nM) for 30 minutes.

Target description

~140 kDa observed. 125 kDa calculated.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhao et al.
Nature communications, 11(1), 3345-3345 (2020-07-06)
Nonsense-mediated mRNA decay (NMD) is an evolutionarily conserved RNA decay mechanism that has emerged as a potent cell-intrinsic restriction mechanism of retroviruses and positive-strand RNA viruses. However, whether NMD is capable of restricting DNA viruses is not known. The DNA
Michiel van Gent et al.
PLoS biology, 19(2), e3001097-e3001097 (2021-02-18)
The oncogenic human herpesviruses Epstein-Barr virus (EBV) and Kaposi's sarcoma-associated herpesvirus (KSHV) are the causative agents of multiple malignancies. A hallmark of herpesviruses is their biphasic life cycle consisting of latent and lytic infection. In this study, we identified that
Noriki Iwai et al.
Nature communications, 15(1), 5765-5765 (2024-07-10)
The ATP-dependent RNA helicase UPF1 plays a crucial role in various mRNA degradation pathways, most importantly in nonsense-mediated mRNA decay (NMD). Here, we show that UPF1 is upregulated during the early stages of B cell development and is important for
Feng Wang et al.
Nature communications, 14(1), 4760-4760 (2023-08-09)
Long-read RNA sequencing (RNA-seq) is a powerful technology for transcriptome analysis, but the relatively low throughput of current long-read sequencing platforms limits transcript coverage. One strategy for overcoming this bottleneck is targeted long-read RNA-seq for preselected gene panels. We present
Oliver Meikar et al.
RNA (New York, N.Y.), 20(4), 483-495 (2014-02-21)
The genome of male germ cells is actively transcribed during spermatogenesis to produce phase-specific protein-coding mRNAs and a considerable amount of different noncoding RNAs. Ribonucleoprotein (RNP) granule-mediated RNA regulation provides a powerful means to secure the quality and correct expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service