Skip to Content
Merck
All Photos(1)

Key Documents

05-822

Sigma-Aldrich

Anti-Rho (-A, -B, -C) Antibody, clone 3L74, rabbit monoclonal

clone 3L74, Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
3L74, monoclonal
application:
WB
species reactivity:
rat, human
technique(s):
western blot: suitable
citations:
5

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3L74, monoclonal

species reactivity

rat, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RHOA(387)

Specificity

Broad species cross-reactivity expected based on sequence homology
Recognizes Rho (-A, -B, -C)

Immunogen

KLH-conjugated, synthetic peptide (SYPDTDVILMCFSIDSPDSLENKK) corresponding to amino acids 73-94 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility

Application

Anti-Rho (-A Antibody, -B, -C), clone 3L74 is an antibody against Rho (-A, -B, -C) for use in WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on modified RIPA lysates from HL-60 cells

Target description

Mr 22kDa

Physical form

Format: Purified
Protein A purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prognostic value of rho GTPases and rho guanine nucleotide dissociation inhibitors in human breast cancers.
Jiang, Wen G, et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 9, 6432-6440 (2003)
D T Denhardt
The Biochemical journal, 318 ( Pt 3), 729-747 (1996-09-15)
The features of three distinct protein phosphorylation cascades in mammalian cells are becoming clear. These signalling pathways link receptor-mediated events at the cell surface or intracellular perturbations such as DNA damage to changes in cytoskeletal structure, vesicle transport and altered
Kyucheol Cho et al.
Journal of cell science, 123(Pt 23), 4128-4144 (2010-11-11)
In common with other p120-catenin subfamily members, Xenopus ARVCF (xARVCF) binds cadherin cytoplasmic domains to enhance cadherin metabolic stability or, when dissociated, modulates Rho-family GTPases. We report here that xARVCF binds and is stabilized by Xenopus KazrinA (xKazrinA), a widely
Stanislav Kholmanskikh et al.
Scientific reports, 12(1), 16493-16493 (2022-10-04)
Regulation of Rho GTPases remains a topic of active investigation as they are essential participants in cell biology and the pathophysiology of many human diseases. Non-degrading ubiquitination (NDU) is a critical regulator of the Ras superfamily, but its relevance to
S Vincent et al.
Molecular and cellular biology, 17(4), 2247-2256 (1997-04-01)
The Ras-related Rho family GTPases mediate signal transduction pathways that regulate a variety of cellular processes. Like Ras, the Rho proteins (which include Rho, Rac, and CDC42) interact directly with protein kinases, which are likely to serve as downstream effector

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service