Skip to Content
Merck
All Photos(3)

Key Documents

05-802R

Sigma-Aldrich

Anti-phospho-Akt (Thr308) Antibody, clone 50-1C-25, rabbit monoclonal

clone NL50, Upstate®, from rabbit

Synonym(s):

Protein kinase B, RAC-alpha serine/threonine-protein kinase, murine thymoma viral (v-akt) oncogene homolog-1, rac protein kinase alpha, v-akt murine thymoma viral oncogene homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NL50, monoclonal

species reactivity

human, bovine, mouse, rabbit, chicken

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr308)

Gene Information

bovine ... Akt1(280991)
human ... AKT1(207)

General description

Akt/PKB is a Ser/Thr kinase and a major known effecter of the PI3 Kinase pathway. It is involved in multiple signaling pathways that relate to many biological processes including metabolism, apoptosis, cell cycle control, angiogenesis, differentiation, and cell growth and proliferation. In mammals, three isoforms of Akt (Akt1/PKCα, Akt2/PKBβ, and Akt3/PKBγ) exists. They exhibit a high degree of homology, but differ slightly in the localization of their regulatory phosphorylation sites. Akt1 is the predominant isoform that is in most tissues and is thought to have a dominant role in growth, survival, embryonic development, and adipocyte differentiation. Akt2 is correlated with the regulation of glucose homeostasis and is the predominant isoform expressed in insulin-responsive tissues. Akt3 is abundant in brain tissue. Each Akt isoform is composed of three functionally distinct regions: an N-terminal Pleckstrin Homology (PH) domain that provides a lipid-binding module, a central catalytic domain containing Thr308, and a C-terminal hydrophobic motif containing Ser473. The activation of AKT is dependent on a dual regulatory mechanism that requires both its translocation to the plasma membrane and dual phosphorylation on Thr308 and Ser473 by PDK1 and the TORC2 complex, respectively.

Specificity

Detects Akt only when phosphorylated on Thr308.

Immunogen

KLH-conjugated synthetic peptide containing the Thr308 sequence and surrounding amino acids of human, mouse, rat, bovine and chicken Akt1.

Application

Detect phospho-Akt (Thr308) using this Anti-phospho-Akt (Thr308) Antibody, clone 50-1C-25 validated for use in WB, IF, IC, ELISA.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
Western Blot Analysis:
This lot detected Akt when phosphorylated on Thr308 at 1:1,000 dilution in PDGF-treated NIH/3T3 lysates via SDS-PAGE and transferred to PVDF (Immobilon-P).

Quality

This lot detected Akt when phosphorylated on Thr308 at 1:1,000 dilution in PDGF-treated NIH/3T3 lysates via SDS-PAGE and transferred to PVDF (Immobilon-P).

Target description

60 kDa

Linkage

Replaces: 05-802

Physical form

Format: Purified
Protein A purified
Protein A purified recombinant rabbit monoclonal supernatant in 0.1M Tris-Glycine (pH7.4) 150mM NaCl, and 0.05% NaN3 with 40% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott Ackler et al.
Oncogene, 21(2), 198-206 (2002-01-23)
AKT1/protein kinase Balpha is a protein-serine/threonine kinase that regulates multiple targets involved in cell survival and cell cycle progression in a variety of cell types including breast cancer cells. To explore the role of Akt1 in mammary gland function and
D R Alessi et al.
The EMBO journal, 15(23), 6541-6551 (1996-12-02)
Insulin activated endogenous protein kinase B alpha (also known as RAC/Akt kinase) activity 12-fold in L6 myotubes, while after transfection into 293 cells PKBalpha was activated 20- and 50-fold in response to insulin and IGF-1 respectively. In both cells, the
Jun Fu et al.
International journal of molecular medicine, 42(3), 1473-1483 (2018-05-31)
The purpose of the present study was to analyze the association between paeonol and the known genes related to gastric cancer (GC) using bioinformatics methods, and to investigate the role of paeonol in the potential impact on the nuclear factor‑κB
Anabel Martínez Báez et al.
Current issues in molecular biology, 45(3), 2296-2308 (2023-03-29)
Insulin signaling plays an important role in the development and progression of cancer since it is involved in proliferation and migration processes. It has been shown that the A isoform of the insulin receptor (IR-A) is often overexpressed, and its
S R James et al.
The Biochemical journal, 315 ( Pt 3), 709-713 (1996-05-01)
Recent evidence has suggested that activation of phosphoinositide 3-kinase (PI 3-kinase) is required for the activation of Akt-1 by growth factors and insulin. Here we demonstrate by two independent methods that Akt-1 from L6 myotubes binds to PtdIns(3,4,5)P3, PtdIns(3,4)P2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service