Skip to Content
Merck
All Photos(1)

Key Documents

05-769

Sigma-Aldrich

Anti-Integrin beta 3 (ITGB3/CD61) Antibody

UPSTATE®, mouse monoclonal, SAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin β3 (CD61) Antibody, clone SAP, clone SAP, Upstate®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SAP, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

Specificity

Integrin β3

Immunogen

Human platelet membrane fractions

Application

Anti-Integrin β3 (CD61) Antibody, clone SAP detects level of Integrin β3 (CD61) & has been published & validated for use in WB, IC, IH.
Research Category
Cell Structure
Research Sub Category
Integrins

Quality

routinely evaluated by immunocytochemistry

Target description

Mr ~115kDa

Linkage

Replaces: 04-1060

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Format: Purified
Protein G Purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Due to license agreement restrictions, this product cannot be purchased for resale.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrin alpha v beta 3 as a therapeutic target for blocking tumor-induced angiogenesis.
Kumar, C Chandra
Current Drug Targets, 4, 123-131 (2003)
Anti-integrin therapy.
Hamm, Christian W
Annual Review of Medicine, 54, 425-435 (2003)
Constanza Garcia-Keller et al.
Biological psychiatry, 86(5), 377-387 (2019-05-28)
Cue-induced relapse to drug use is a primary symptom of cocaine addiction. Cue-induced transient excitatory synaptic potentiation (t-SP) induced in the nucleus accumbens mediates cued cocaine seeking in rat models of relapse. Cue-induced t-SP depends on extracellular signaling by matrix

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service