Skip to Content
Merck
All Photos(1)

Key Documents

05-615

Sigma-Aldrich

Anti-SUV39H1 Histone Methyltransferase antibody

UPSTATE®, mouse monoclonal, MG44

Synonym(s):

KMT1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-SUV39H1 Antibody, clone MG44, ascites fluid, clone MG44, Upstate®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MG44, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SUV39H1(6839)

General description

Post -translational modification of histones, such as phosphorylation, acetylation, and methylation, are crucial for chromatin remodeling. SUV39H1 and Suv39h1 are the human and mouse homologs of Drosophila Su(var)3-9 protein, respectively. These proteins selectively methylate histone H3 at lysine 9 creating a high-affinity binding site for the HP1 proteins (heterochromatin protein 1). HP1 proteins are known to interact with transcription suppressor proteins, suggesting that the interactions with SUV39H1 and methylated histone H3 mediate a silence effect on the transcription of target genes. Furthermore, over-expression of SUV39H1 in HeLa cells causes a redistribution of endogenous HP1 proteins and growth retardation, suggesting that SUV39H1-mediated modulation of heterochromatin can impair cell cycle progression.

Specificity

Recognizes SUV39H1.

Immunogen

MBP fusion protein corresponding to residues 1-412 of human SUV39H1

Application

Anti-SUV39H1 Antibody, clone MG44 is a Mouse Monoclonal Antibody for detection of SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1 & has been validated in ChIP, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology

Quality

routinely evaluated by immunoblot on U2OS cells lysed in RIPA with 0.1% SDS and sonicated

Target description

50 kDa

Linkage

Replaces: MABE552

Physical form

Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
U2OS cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binbin Liu et al.
Nucleic acids research, 47(1), 283-298 (2018-11-09)
Various stresses increase disease susceptibility and accelerate aging, and increasing evidence suggests that these effects can be transmitted over generation. Epidemiological studies suggest that stressors experienced by parents affect the longevity of their offspring, possibly by regulating telomere dynamics. Telomeres
Whitney L Johnson et al.
eLife, 6 (2017-08-02)
Heterochromatin formed by the SUV39 histone methyltransferases represses transcription from repetitive DNA sequences and ensures genomic stability. How SUV39 enzymes localize to their target genomic loci remains unclear. Here, we demonstrate that chromatin-associated RNA contributes to the stable association of
Luke Gaughan et al.
The Journal of biological chemistry, 277(29), 25904-25913 (2002-05-08)
The androgen receptor (AR), a member of the nuclear hormone receptor superfamily, is thought to play an important role in the development of prostate cancer. The AR is a hormone-dependent transcription factor that activates expression of numerous androgen-responsive genes. Histone
Liping Deng et al.
Nature communications, 10(1), 3329-3329 (2019-07-28)
DiGeorge syndrome critical region 8 (DGCR8) is a critical component of the canonical microprocessor complex for microRNA biogenesis. However, the non-canonical functions of DGCR8 have not been studied. Here, we demonstrate that DGCR8 plays an important role in maintaining heterochromatin
Suv39h1 mediates AP-2?-dependent inhibition of C/EBP? expression during adipogenesis.
Zhang, ZC; Liu, Y; Li, SF; Guo, L; Zhao, Y; Qian, SW; Wen, B; Tang, QQ; Li, X
Molecular and cellular biology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service