Skip to Content
Merck
All Photos(3)

Key Documents

05-495

Sigma-Aldrich

Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 8-5-2

clone 8-5-2, Upstate®, from mouse

Synonym(s):

signal transducer and activator of transcription 5A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8-5-2, monoclonal

species reactivity

mouse, sheep, human, rat, bovine

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr694/pTyr699)

Gene Information

human ... STAT5A(6776)

General description

STAT (Signal Transducers and Activators of Transcription) are a family of conserved transcription factors that transduce high-fidelity signals for the cytokine family of ligands and receptors as the principle substrates of JAK kinases (1,2). In resting cells, the STATs exist in an inactive state in the cytoplasm until activated. Activators include cytokines, growth factors, and some peptides. These activate JAK kinases, which when activated, bind to the STAT SH2 (Src homology domain-2) domain resulting in phosphorylation of STAT (1). Upon activation, STAT1, -3, -4, -5A, and -5B all form homodimers. STAT1 and STAT2, as well as STAT1 and STAT3 form heterodimers, and several STATs form tetramers, depending on the variables of the activating ligand (1). After dimerization, the STATs accumulate in the nucleus by way of nuclear transport (1). STAT5A and STAT5B appear to be most important to the function of growth hormone and prolactin and play critical roles in milk production (1,2,3). The phosphorylation of Tyrosine 694/699 is a hallmark of activated STAT5.

Specificity

Specific for the phosphorylated tyrosine residue 694 of STAT5A (94kDa) and 699 of STAT5B (92kDa). Unidentified proteins may be detected in some lysates.

Immunogen

Epitope: Tyr694/699
KLH-conjugated, synthetic phosphopeptide (KAVDG[pY]VKPQIK) corresponding to amino acids 689-700 of STAT5A and 694-705 of STAT5B. Clone 8-5-2.

Application

Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 8-5-2 is a mouse monoclonal antibody for detection of phospho-STAT5A/B (Tyr694/699) also known as signal transducer & activator of transcription 5A/B & has been validated in WB.

Quality

Routinely evaluated by Western Blot on 3T3/A31 cells stimulated with PDGF or HeLa cells stimulated with interferon-α.

Western Blot Analysis: 0.5-2 μg/mL of this lot detected phosphorylated STAT5A/B in lysates from 3T3/NIH cells stimulated with PDGF. 0.5-2 μg/mL of a previous lot detected phosphorylated STAT5A/B in lysates from 3T3/A31 cells stimulated with PDGF and from HeLa cells stimulated with interferon-a (IFN-a).

Target description

STAT5A (94 kDa) and STAT5B (92 kDa)

Physical form

Format: Purified
Purified mouse monoclonal IgG in buffer containing PBS. Frozen solution at -20°C.

Analysis Note

Control
IFN-α induced HeLa cells or GM-CSF treated TF-1 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Up-regulation of SLAP in FLI-1-transformed erythroblasts interferes with EpoR signaling.
Lebigot, I; Gardellin, P; Lefebvre, L; Beug, H; Ghysdael, J; Quang, CT
Blood null
Forced involution of the functionally differentiated mammary gland by overexpression of the pro-apoptotic protein bax.
Edmund B Rucker,Amber N Hale,David C Durtschi,Kazuhito Sakamoto,Kay-Uwe Wagner
Genesis (2000)
T M Schnöder et al.
Cell death and differentiation, 22(6), 974-985 (2014-11-15)
Erythropoiesis is a tightly regulated process. Development of red blood cells occurs through differentiation of hematopoietic stem cells (HSCs) into more committed progenitors and finally into erythrocytes. Binding of erythropoietin (Epo) to its receptor (EpoR) is required for erythropoiesis as
IL-7 activates the phosphatidylinositol 3-kinase/AKT pathway in normal human thymocytes but not normal human B cell precursors.
Sonja E Johnson, Nisha Shah, Anna A Bajer, Tucker W LeBien
Journal of immunology (Baltimore, Md. : 1950) (1950)
Xianping Shi et al.
Scientific reports, 5, 9694-9694 (2015-04-09)
Resistance to chemotherapy is a great challenge to improving the survival of patients with diffuse large B-cell lymphoma (DLBCL), especially those with activated B-cell-like DLBCL (ABC-DLBCL). Therefore it is urgent to search for novel agents for the treatment of DLBCL.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service