Skip to Content
Merck
All Photos(1)

Documents

05-338

Sigma-Aldrich

Anti-Fas Antibody (human, neutralizing), clone ZB4

clone ZB4, Upstate®, from mouse

Synonym(s):

APO-1 cell surface antigen, Apo-1 antigen, Apoptosis-mediating surface antigen FAS, CD95 antigen, FASLG receptor, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, apoptosis antigen 1, tumor necrosis factor receptor superfamily member 6, tu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ZB4, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
neutralization: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FAS(355)

General description

Fas (also known as Apo-1 and CD95) is a 40-50 kDa cell surface glycoprotein and a member of the TNF receptor family. Fas is an apotosis signalling molecule that forms the death inducing signalling complex (DISC) upon binding by the Fas ligand trimer on an adjacent cell. The Fas receptor also trimerizes, after which it is internalized and can then bind to the adaptor molecule FADD. FADD facilitates binding of caspase-8. which is activated to induce caspase-8 mediated apoptosis. Fas also plays a role in T cell mediated cytotoxicity and T cell development. Fas is found in the thymus, liver, heart, and ovary, where it is expressed on a minority of resting T and B cells and monocytes. The antagonistic ZB4 clone anti-Fas monoclonal has been used to block the apoptosis-inducing activity of the Anti-Fas CH-11 clone in multiple publications.
This product does not induce apoptosis in cell culture.

Specificity

This antibody does not recognize TNF and mouse Fas.
This antibody recognizes the human cell surface antigen Fas expressed in various human cells, including myeloid cells, T lympho-blastoid cells and diploid fibroblasts. It does not induce apoptosis in cell culture.

Immunogen

Recombinant human Fas.

Application

Anti-Fas Antibody (human, neutralizing), clone ZB4 detects level of Fas & has been published & validated for use in WB, FC, NEUT.
Flow Cytometry: A previous lot of this antibody used 5-20 μg/mL. Western Blot: A previous lot of this antibody used 5 μg/mL.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by neutralization of apoptosis induced by anti-Fas (human, activating), clone CH11 (Catalog # 05-201) treatment of human Jurkat cells.

Neutralization: 10-500 ng/mL of this antibody was incubated for one hour with human Fas-transfected cells and inhibited the anti-Fas (human, activating), clone CH11 (Catalog # 05-201). Cell viability was assessed using a WST-1 assay

Target description

45 kDa

Physical form

Ammonium sulfate and purified mouse monoclonal IgG1 in PBS containing 50% glycerol. Store at -20°C.
Format: Purified
Protein A purified

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
Various human cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divya Kesanakurti et al.
PloS one, 6(5), e19341-e19341 (2011-05-17)
Abrogation of apoptosis for prolonged cell survival is essential in cancer progression. In our previous studies, we showed the MMP-2 downregulation induced apoptosis in cancer cell lines. Here, we attempt to investigate the exact molecular mechanism of how MMP-2 depletion
Stimulated human lamina propria T cells manifest enhanced Fas-mediated apoptosis
Boirivant, M., et al
The Journal of Clinical Investigation, 98, 2616-2622 (1996)
Networked T cell death following macrophage infection by Mycobacterium tuberculosis.
Macdonald, SH; Woodward, E; Coleman, MM; Dorris, ER; Nadarajan, P; Chew, WM; McLaughlin et al.
Testing null
E Lacana et al.
Nature medicine, 5(5), 542-547 (1999-05-06)
Programmed cell death is a process required for the normal development of an organism. One of the best understood apoptotic pathways occurs in T lymphocytes and is mediated by Fas/Fas ligand (FasL) interaction. During studies of apoptosis induced by T
Molecular determinants of apoptosis induced by the cytotoxic ribonuclease onconase: evidence for cytotoxic mechanisms different from inhibition of protein synthesis.
M S Iordanov, O P Ryabinina, J Wong, T H Dinh, D L Newton, S M Rybak, B E Magun
Cancer Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service