Skip to Content
Merck
All Photos(1)

Key Documents

05-1250

Sigma-Aldrich

Anti-Trimethyl Histone H3(Lys9) Antibody, clone CMA308

clone CMA308, from mouse

Synonym(s):

H3K9me3, Histone H3 (tri methyl K9), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CMA308, monoclonal

species reactivity

vertebrates, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
multiplexing: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys9)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone proteins are highly post-translationally modified however Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Trimethylation of histone H3 on Lys9 (H3K9me3) is one of the most highly studied epigenetic marks. H3K9me3 functions in the repression of euchromatic genes, and in epigenetic control of heterochromatin assembly, most likely by acting as a recognition motif for the binding of chromatin-associated proteins, such as Swi6 or HP1α. The enzymes responsible for H3K9 trimethylation are SUV39H1 and SUV39H2.

Specificity

Broad species cross-reactivity is expected, based on sequence homology.
This antibody recognizes Histone H3 trimethylated at Lys9. Some cross-reactivity with dimethyl Lys9 is detected at higher concentrations, but the difference in specificity is >27-fold1. Phosphorylation of Ser10 interferes with antibody binding to trimethyl Lys91.

Immunogen

Epitope: Trimethyl Lys9
Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, trimethylated on Lys9, conjugated to KLH.

Application

Anti-Trimethyl Histone H3(Lys9) Antibody, clone CMA308 is a mouse monoclonal antibody for detection of Trimethyl Histone H3(Lys9) also known as H3K9me3, Histone H3 (tri methyl K9) & has been validated in WB, ELISA, ICC, IP, Mplex.
ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA.1

Immunoprecipitation:
This antibody has been shown by an outside laboratory to be suitable for IP.1

Immunocytochemistry:
This antibody has been shown by an outside laboratory to be suitable for immunocytochemistry.1
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Western Blot Analysis:
A 0.5 – 2 μg/mL dilution of this antibody detected histone H3 in 10 μg of HeLa acid extract.

Target description

~17 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt. For maximum recovery of product, centrifuge the vial prior to removing the cap. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa Acid extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Gonzalez-Munoz et al.
Scientific reports, 9(1), 8632-8632 (2019-06-16)
Mouse and cell-based studies have shown that macroH2A histone variants predominantly associate with heterochromatin. Functional studies found that macroH2As are involved in gene repression, inhibiting the acquisition of pluripotency and preserving cell differentiation. However, only a few studies have analysed
Toyonori Sakata et al.
Methods in molecular biology (Clifton, N.J.), 1515, 257-271 (2016-11-01)
ChIP-seq, or chromatin immunoprecipitation combined with massively parallel DNA sequencing, is a powerful technique to investigate in vivo protein-DNA interactions on a genome-wide scale at high resolution. Here we describe a ChIP-seq protocol optimized for analysis of condensin I complex
Sebastian Oeck et al.
Scientific reports, 9(1), 3148-3148 (2019-03-01)
DNA- and histone-related research frequently comprises the quantitative analysis of protein modifications, such as histone phosphorylation. Analysis of accumulation and disappearance of protein foci are used to monitor DNA damage and repair kinetics. If the protein of interest doesn't accumulate
Hanqing Zhao et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(4), 918-929 (2015-02-28)
In this study, we attempted to uncover the functional impact of microRNA-22 (miR-22) and its target gene in smooth muscle cell (SMC) differentiation and delineate the molecular mechanism involved. miR-22 was found to be significantly upregulated during SMC differentiation from
Dijana Lađinović et al.
Nucleus (Austin, Tex.), 8(5), 563-572 (2017-08-18)
Histone modifications have a profound impact on the chromatin structure and gene expression and their correct establishment and recognition is essential for correct cell functioning. Malfunction of histone modifying proteins is associated with developmental defects and diseases and detailed characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service