Skip to Content
Merck
All Photos(1)

Key Documents

V-016

Supelco

Nicotinamide (Vitamin B3) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Nicotinamide, Vitamin B3, Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

128-131 °C (lit.)

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide is also known as niacinamide or nicotinic acid amide. It is the water-soluble form of Vitamin B3 found in plant and animal foods and mostly used as a dietary supplement.

Application

This certified solution standard is suitable for a variety of testing applications including quality control testing by HPLC for pharmaceutical preparations or dietary supplements.
Nicotinamide may be used as an analytical reference standard for the determination of the analyte in energy drinks by planar chromatography-multiple detection with confirmation by electrospray ionization mass spectrometry[1] and in pharmaceutical formulations, by UV-visible spectrophotometry and multivariate analysis.[2]

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of riboflavin, pyridoxine, nicotinamide, caffeine and taurine in energy drinks by planar chromatography-multiple detection with confirmation by electrospray ionization mass spectrometry
Aranda M and Morlock G
Journal of Chromatography A, 1131(1-2), 253-260 (2006)
Simultaneous determination and classification of riboflavin, thiamine, nicotinamide and pyridoxine in pharmaceutical formulations, by UV-visible spectrophotometry and multivariate analysis
Lopez-de-Alba LP, et al.
Journal of the Brazilian Chemical Society, 17(4), 715-722 (2006)
Takeshi Ueda et al.
Japanese journal of clinical oncology, 43(6), 616-628 (2013-05-01)
Axitinib is a potent and selective second-generation inhibitor of vascular endothelial growth factor receptors 1, 2 and 3. The efficacy and safety of axitinib in Japanese patients with metastatic renal cell carcinoma were evaluated. A subgroup analysis was conducted in
J Gore et al.
Journal of economic entomology, 106(3), 1430-1439 (2013-07-20)
Cotton aphid, Aphis gossypii Glover, has a history of developing resistance to novel insecticides. A program is needed to monitor cotton aphid susceptibility to new insecticides. Concentration-mortality bioassays were conducted from 2008 to 2011 to monitor the susceptibility of cotton
Ning Jia et al.
Anticancer research, 33(6), 2797-2800 (2013-06-12)
Post-transplant hepatocellular carcinoma recurrence has been reported to be between 15-18% and is higher among patients with high-risk features (bilobar tumor, macrovascular invasion, or multifocality). There are no known treatments which reduce risk of recurrence post-transplant. Sorafenib is currently approved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service