Skip to Content
Merck
All Photos(1)

Key Documents

S-046

Supelco

JWH-250 4-Hydroxypentyl metabolite

ampule of 1 mL, certified reference material, 100 μg/mL in methanol, Cerilliant®

Synonym(s):

JWH-250 4-Hydroxypentyl metabolite solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO3
Molecular Weight:
351.44
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COc1ccccc1CC(=O)c2cn(CCCC(C)O)c3ccccc23

InChI

1S/C22H25NO3/c1-16(24)8-7-13-23-15-19(18-10-4-5-11-20(18)23)21(25)14-17-9-3-6-12-22(17)26-2/h3-6,9-12,15-16,24H,7-8,13-14H2,1-2H3

InChI key

GMXLLEJMDHWZNM-UHFFFAOYSA-N

General description

A Certified Spiking Solution® of a JWH-250 biomarker often referred to as the omega-1 hydroxy metabolite of JWH-250. This monohydroxy analog is a primary urinary metabolite of JWH-250, a synthetic cannabinoid found in blends of the herbal mixture Spice.

Application


  • Doping control analysis of four JWH-250 metabolites in equine urine by liquid chromatography-tandem mass spectrometry: This study examines the detection of JWH-250 metabolites in equine urine, contributing to doping control efforts in sports (You et al., 2019).

  • Urinary prevalence, metabolite detection rates, temporal patterns and evaluation of suitable LC-MS/MS targets to document synthetic cannabinoid intake in US military urine specimens: This study identifies the prevalence and patterns of synthetic cannabinoid use among military personnel through urinary analysis (Wohlfarth et al., 2015).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service