Skip to Content
Merck
All Photos(1)

Key Documents

R-024

Supelco

Remifentanil hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O5 · HCl
CAS Number:
Molecular Weight:
412.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[Cl-].N1(CCC(CC1)(N(c2ccccc2)C(=O)CC)C(=O)OC)CCC(=O)OC.[H+]

InChI

1S/C20H28N2O5.ClH/c1-4-17(23)22(16-8-6-5-7-9-16)20(19(25)27-3)11-14-21(15-12-20)13-10-18(24)26-2;/h5-9H,4,10-15H2,1-3H3;1H

InChI key

WFBMIPUMYUHANP-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

Remifentanil is a potent, short-acting, synthetic opioid analgesic drug. It is used in combination with other opioids and analgesics to induce or supplement general anaesthesia. This new certified reference solution is suitable for a variety of mass spectrometry-based testing applications including clinical toxicology, forensic analysis, workplace drug testing and pharmaceutical research.

Application

  • Analgesic efficacy studies using Remifentanil hydrochloride: A comprehensive analysis focused on the simultaneous determination of 14 analgesics, including Remifentanil hydrochloride, in postoperative solutions using HPLC-DAD and LC-MS/MS methodologies, highlighting its role in effective pain management protocols (Yao et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service