Skip to Content
Merck
All Photos(1)

Documents

P-055

Supelco

Propranolol hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO2·HCl
CAS Number:
Molecular Weight:
295.80
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CC(C)NCC(O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H

InChI key

ZMRUPTIKESYGQW-UHFFFAOYSA-N

Gene Information

General description

A certified reference solution suitable for use as starting material in calibrators and controls for LC/MS or GC/MS testing methods in clinical and diagnostic testing, urine drug testing, forensic analysis, or clinical toxicology. Propranolol is a sympatholytic non-selective beta blocker used to treat a variety of medical conditions including hypertension, anxiety and panic. The drug is sold under numerous brand names such as Inderal®, Avlocardyl, Deralin, and Dociton.

Application

<ul>
<li><strong>Propranolol hydrochloride solution for infantile hemangioma:</strong>Liposomal formulations of propranolol hydrochloride have been developed for the treatment of infantile hemangioma, offering a targeted therapeutic approach in pediatric dermatology (Nifli et al., 2024).</li>
<li><strong>Propranolol hydrochloride:</strong> Research on the percutaneous absorption of solvent-deposited propranolol hydrochloride across different skin layers provides insights into its properties like it can be used as a function of dose and ionization state and as a calibration standard (Tonnis et al., 2024).</li>
</ul>

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Inderal is a registered trademark of Cranford Pharmaceuticals, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip M Short et al.
American journal of respiratory and critical care medicine, 187(12), 1308-1314 (2013-04-19)
Unblinded studies have shown improvements in airway hyperresponsiveness with chronic nadolol in steroid-naive patients with asthma. To assess the effects of chronic nonselective β-blockade as add-on to inhaled corticosteroids (ICS) in patients with asthma. A double-blind randomized placebo-controlled crossover trial
Jun Zhang et al.
Rapid communications in mass spectrometry : RCM, 27(7), 731-737 (2013-03-16)
Multiplexed liquid chromatography (LC) coupled with multiple-injection-chromatogram acquisition has emerged as the method of choice for high-speed discovery bioanalysis, because it significantly reduces injection-to-injection cycle time while maintaining the chromatography quality. Historically, systems utilizing this approach had been custom built
[Propanolol effective treatment for laryngo-tracheal hemangioma].
Markus Granholm et al.
Lakartidningen, 110(19-20), 947-948 (2013-06-12)
What doesn't kill may not make you stronger. β-blockers for asthma.
Shamsah Kazani et al.
American journal of respiratory and critical care medicine, 187(12), 1281-1281 (2013-06-19)
Bojko Bjelakovic et al.
Drug design, development and therapy, 7, 127-129 (2013-03-08)
We hypothesized that some characteristics of beta-blockers, including negative inotropic, peripheral vasoconstrictor, and antiangiogenic effects, might be potentially useful in treating children with epistaxis. From June 2010 to March 2012, a total of seven children with recurrent primary epistaxis resistant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service